Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00572430_m1
          See other CNKSR2 GE Assays ›
          Gene Symbol:
          Cnksr2
          Gene Name:
          connector enhancer of kinase suppressor of Ras 2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.X: 39711042 - 39953860 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00572430_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cnksr2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001113366.1 NP_001106837.1 8 - 9 982 106
          NM_021686.3 NP_067718.1 8 - 9 982 106
          XM_008773224.1 XP_008771446.1 8 - 9 1141 106
          GenBank mRNA AF102853.1 - 8 - 9 809 106
          AF102854.1 - 8 - 9 1271 106

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0X00000382.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cnksr2

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of signal transduction
          protein binding
          identical protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P178576 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P196440 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 136250 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 396798 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 600103 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 957863 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 136631 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 418325 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 731996 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 682273 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 45541 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 626243 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 715749 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 878798 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 43634 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 905714 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 416721 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 116114 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 567805 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 918642 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 944406 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 158991 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 757715 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 200641 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 884336 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 807266 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 465283 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 67456 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 318819 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 917186 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 907901 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 18046 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 412728 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 215431 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 218572 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 778145 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 184407 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 836970 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 56238 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 396844 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 60149 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 409122 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 666645 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 317465 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 871475 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 256662 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 556618 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 359824 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 821853 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 424379 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 489177 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 533269 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 756456 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 292704 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 894566 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 705717 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 54093 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 746589 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 927524 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 587713 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 402891 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 664070 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 739313 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 456406 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 674117 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 227514 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 686615 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 649115 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 189084 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 446019 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 704411 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 501502 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 690380 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 881743 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 222069 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 751657 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 299636 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 4333 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 667116 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 6334 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 293002 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 239289 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 766495 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 808642 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 82950 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 864770 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 946039 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 255021 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 407205 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 502168 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 18989 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 246010 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 355679 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 789896 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 124832 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 286374 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 701448 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 194207 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 379725 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 520230 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 711862 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 630598 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 53299 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 301758 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 13657 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 791961 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 476658 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 512053 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 652841 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 794478 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 460748 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 457165 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 209848 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 8112 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 320466 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 927899 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 390986 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 375565 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 235506 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 160528 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 846059 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 195297 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 721277 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 229472 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 317525 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 783131 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 583384 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 511153 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 725432 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 55109 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 665277 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 800445 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 108425 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 231115 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 34126 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 180953 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 879407 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 178914 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 501574 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 643510 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 553398 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 595412 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 729185 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 461617 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 774587 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 848675 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 451377 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 678604 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 647947 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 211756 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 770449 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 84567 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 696401 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 268315 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 146539 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 968434 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 666979 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 961122 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 918792 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 11833 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 498379 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 319763 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 537452 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 108063 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 463405 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 631980 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 803711 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 875984 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 578124 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 822493 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 56006 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 37410 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 57282 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 155334 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 44203 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 119806 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 113546 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 172838 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 156938 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 137720 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 136822 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 55210 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 43780 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 145252 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 178457 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 103761 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 93596 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 153067 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 73608 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 58894 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 167706 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 177957 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 12943 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 144984 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 41219 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 141512 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 55498 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 52390 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 140596 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 149462 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P196440 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P178576 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P178576 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P196440 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P196440 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P178576 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P178576 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 88227 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 61763 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 39096 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 86075 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 81217 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 67589 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 12886 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 34985 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 70219 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 73836 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 34277 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 29851 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 4134 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 22120 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 93248 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 15494 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 675 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10933611 - -
          Affymetrix Rat Gene 1.0 ST Array 10933624 - -
          Affymetrix Rat Gene 1.0 ST Array 10933620 - -
          Affymetrix Rat Gene 1.0 ST Array 10933632 - -
          Affymetrix Rat Gene 1.0 ST Array 10933631 - -
          Affymetrix Rat Gene 1.0 ST Array 10933618 - -
          Affymetrix Rat Gene 1.0 ST Array 10933626 - -
          Affymetrix Rat Gene 1.0 ST Array 10933629 - -
          Affymetrix Rat Gene 1.0 ST Array 10933621 - -
          Affymetrix Rat Gene 1.0 ST Array 10933610 - -
          Affymetrix Rat Gene 1.0 ST Array 10933613 - -
          Affymetrix Rat Gene 1.0 ST Array 10933633 - -
          Affymetrix Rat Gene 1.0 ST Array 10933622 - -
          Affymetrix Rat Gene 1.0 ST Array 10933615 - -
          Affymetrix Rat Gene 1.0 ST Array 10933614 - -
          Affymetrix Rat Gene 1.0 ST Array 10933623 - -
          Affymetrix Rat Gene 1.0 ST Array 10933617 - -
          Affymetrix Rat Gene 1.0 ST Array 10933612 - -
          Affymetrix Rat Gene 1.0 ST Array 10933627 - -
          Affymetrix Rat Gene 1.0 ST Array 10933625 - -
          Affymetrix Rat Gene 1.0 ST Array 10933616 - -
          Affymetrix Rat Gene 1.0 ST Array 10933630 - -
          Affymetrix Rat Gene 1.0 ST Array 10933619 - -
          Affymetrix Rat Gene 1.0 ST Array 10933628 - -
          Affymetrix Rat Gene 1.1 ST Array 10933609 - -
          Rat Genome 230 2.0 Array 1370668_PM_a_at - -
          Rat Genome 230 2.0 Array 1369731_PM_at - -
          Rat Genome 230 2.0 Array 1369731_at - -
          Rat Genome 230 2.0 Array 1370668_a_at - -
          Affymetrix HT Rat Focus Array 1369731_PM_at - -
          Affymetrix HT Rat Focus Array 1370668_a_at - -
          Affymetrix HT Rat Focus Array 1370668_PM_a_at - -
          Affymetrix HT Rat Focus Array 1369731_at - -
          Affymetrix Rat Clariom S Assay PSR0X00002690.rn.1 TC0X00000382.rn.1 RTA TC
          Rat Expression Set 230 1370668_a_at - -
          Rat Expression Set 230 1369731_PM_at - -
          Rat Expression Set 230 1369731_at - -
          Rat Expression Set 230 1370668_PM_a_at - -
          Affymetrix Rat Gene 1.0 ST Array 10933609 - -
          Rat Genome U34 Set AF102853_g_at - -
          Rat Genome U34 Set AF102853_at - -
          Rat Genome U34 Set AF102854_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21520979 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1348904 - -
          RatRef-12 Expression BeadChip ILMN_59228 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0