Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00573537_m1
          See other UNC5A GE Assays ›
          Gene Symbol:
          Unc5a
          Gene Name:
          unc-5 homolog A (C. elegans)
          Gene Aliases:
          Unc5h1
          Chromosome Location:
          Chr.17: 10152949 - 10208599 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00573537_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Unc5a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_022206.1 NP_071542.1 2 - 3 293 73
          XM_008771505.1 XP_008769727.1 2 - 3 532 73
          GenBank mRNA U87305.1 - 2 - 3 293 73

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1700001335.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Unc5a

          Gene Ontology Categories:

          Function(s) Process(es)

          apoptotic process
          signal transduction
          axon guidance
          anterior/posterior axon guidance
          netrin-activated signaling pathway
          netrin receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P040067 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 529366 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 64751 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 229199 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 339266 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 595592 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 880922 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 211407 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 546936 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 385465 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 200169 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 162204 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 105100 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 689811 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 260341 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 954150 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 363648 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 848370 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 417074 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 115258 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 851321 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 192971 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 226846 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 351672 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 522330 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 365679 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 386215 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 131411 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 410583 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 483504 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 301938 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 970932 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 284492 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 698138 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 605994 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 411767 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 716992 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 435008 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 749786 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 468014 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 266384 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 918791 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 736491 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 874373 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 128885 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 17066 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 20224 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 100571 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 172150 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 7405 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 148352 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 39297 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P040067 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P040067 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P040067 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P558378 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 2125 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 19911 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 11025 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 28614 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 27118 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10797357 - -
          Affymetrix Rat Gene 1.0 ST Array 10797364 - -
          Affymetrix Rat Gene 1.0 ST Array 10797361 - -
          Affymetrix Rat Gene 1.0 ST Array 10797358 - -
          Affymetrix Rat Gene 1.0 ST Array 10797367 - -
          Affymetrix Rat Gene 1.0 ST Array 10797355 - -
          Affymetrix Rat Gene 1.0 ST Array 10797353 - -
          Affymetrix Rat Gene 1.0 ST Array 10797365 - -
          Affymetrix Rat Gene 1.0 ST Array 10797352 - -
          Affymetrix Rat Gene 1.0 ST Array 10797366 - -
          Affymetrix Rat Gene 1.0 ST Array 10797363 - -
          Affymetrix Rat Gene 1.0 ST Array 10797362 - -
          Affymetrix Rat Gene 1.0 ST Array 10797360 - -
          Affymetrix Rat Gene 1.0 ST Array 10797356 - -
          Affymetrix Rat Gene 1.0 ST Array 10797359 - -
          Affymetrix Rat Gene 1.0 ST Array 10797354 - -
          Affymetrix Rat Gene 1.1 ST Array 10797351 - -
          Rat Genome 230 2.0 Array 1369394_at - -
          Rat Genome 230 2.0 Array 1369394_PM_at - -
          Affymetrix HT Rat Focus Array 1369394_at - -
          Affymetrix HT Rat Focus Array 1369394_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1700009317.rn.1 TC1700001335.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700009318.rn.1 TC1700001335.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700009315.rn.1 TC1700001335.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700009316.rn.1 TC1700001335.rn.1 RTA TC
          Rat Expression Set 230 1369394_PM_at - -
          Rat Expression Set 230 1369394_at - -
          Affymetrix Rat Gene 1.0 ST Array 10797351 - -
          Rat Genome U34 Set U87305_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20840317 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1370320 - -
          RatRef-12 Expression BeadChip ILMN_49385 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7699c4d8ff-7jgns:80/100.66.130.62:80.
          git-commit: c11338b6b8d815146c5ef25758b63739976b60cb
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline