Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00574337_m1
          See other CAP1 GE Assays ›
          Gene Symbol:
          Cap1
          Gene Name:
          CAP, adenylate cyclase-associated protein 1 (yeast)
          Gene Aliases:
          Mch1
          Chromosome Location:
          Chr.5: 140559191 - 140585489 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00574337_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cap1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_022383.2 NP_071778.2 12 - 13 1461 112
          XM_006238830.2 XP_006238892.1 12 - 13 1561 112
          XM_006238831.2 XP_006238893.1 12 - 13 1558 112
          XM_006238832.2 XP_006238894.1 12 - 13 1539 112
          GenBank mRNA BC089801.1 - 12 - 13 1461 112
          L11930.1 - 11 - 12 1363 112

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0500003539.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cap1

          Gene Ontology Categories:

          Function(s) Process(es)

          cell morphogenesis
          ameboidal-type cell migration
          receptor-mediated endocytosis
          biological_process
          actin cytoskeleton organization
          molecular_function
          actin binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P12212 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 840281 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 650216 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 529057 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 401971 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 562570 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 602365 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 663554 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 503144 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 645773 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 607281 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 668728 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 420613 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 943331 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 931169 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 910890 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 703677 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 312385 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 200718 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 263124 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 689573 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 786526 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 95870 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 163613 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 3363 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P12212 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P12212 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P12212 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P12212 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 77749 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 6618 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 82988 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10879514 - -
          Affymetrix Rat Gene 1.0 ST Array 10879506 - -
          Affymetrix Rat Gene 1.0 ST Array 10879513 - -
          Affymetrix Rat Gene 1.0 ST Array 10879508 - -
          Affymetrix Rat Gene 1.0 ST Array 10879504 - -
          Affymetrix Rat Gene 1.0 ST Array 10879503 - -
          Affymetrix Rat Gene 1.0 ST Array 10879510 - -
          Affymetrix Rat Gene 1.0 ST Array 10879505 - -
          Affymetrix Rat Gene 1.0 ST Array 10879515 - -
          Affymetrix Rat Gene 1.0 ST Array 10879509 - -
          Affymetrix Rat Gene 1.0 ST Array 10879512 - -
          Affymetrix Rat Gene 1.0 ST Array 10879511 - -
          Affymetrix Rat Gene 1.0 ST Array 10879507 - -
          Affymetrix Rat Gene 1.1 ST Array 10879502 - -
          Rat Genome 230 2.0 Array 1368809_PM_at - -
          Rat Genome 230 2.0 Array 1368808_at - -
          Rat Genome 230 2.0 Array 1368808_PM_at - -
          Rat Genome 230 2.0 Array 1368809_at - -
          Affymetrix HT Rat Focus Array 1368808_at - -
          Affymetrix HT Rat Focus Array 1368808_PM_at - -
          Affymetrix HT Rat Focus Array 1368809_PM_at - -
          Affymetrix HT Rat Focus Array 1368809_at - -
          Affymetrix Rat Clariom S Assay PSR0500026226.rn.1 TC0500003539.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500026223.rn.1 TC0500003539.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500026227.rn.1 TC0500003539.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500026221.rn.1 TC0500003539.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500026222.rn.1 TC0500003539.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500026225.rn.1 TC0500003539.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500026228.rn.1 TC0500003539.rn.1 RTA TC
          Rat Expression Set 230 1368809_at - -
          Rat Expression Set 230 1368808_at - -
          Rat Expression Set 230 1368808_PM_at - -
          Rat Expression Set 230 1368809_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10879502 - -
          Rat Genome U34 Set L11930_at - -
          Rat Genome U34 Set rc_AA996997_f_at - -
          Rat Genome U34 Set rc_AA850040_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21960020 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1651181 - -
          RatRef-12 Expression BeadChip ILMN_58645 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5777fb6d44-4pg6x:80/100.66.134.75:80.
          git-commit: 3a4c0ffdaddae765c0fdfab5c280296089abf14d
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0