Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00575566_m1
          See other MBL2 GE Assays ›
          Gene Symbol:
          Mbl2 LOC100911854
          Gene Name:
          mannose-binding lectin (protein C) 2
          mannose-binding protein C-like
          Gene Aliases:
          Ab2-001, Ab2-011
          -
          Chromosome Location:
          Chr.1: 248435069 - 248442669 on Build Rnor_6.0
          Chromosome Location:
          Chr.1: 248723370 - 248730969 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Rn00575566_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mbl2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_022704.2 NP_073195.2 3 - 4 374 97
          XM_006231249.2 XP_006231311.1 5 - 6 650 97
          XM_006231250.2 XP_006231312.1 5 - 6 570 97
          GenBank mRNA AY325174.1 - 404 97
          AY325178.1 - 404 97
          FQ210356.1 - 5 - 6 559 97
          M14103.1 - 5 - 6 544 97
          X05023.1 - 3 - 4 374 97
          Gene Symbol: LOC100911854
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_003749087.3 XP_003749135.1 650 97
          XM_003749088.3 XP_003749136.1 570 97

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100003465.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Mbl2
          Click here to find siRNAs for Gene LOC100911854

          Gene Ontology Categories:

          Function(s) Process(es)

          complement activation, lectin pathway
          complement activation, classical pathway
          positive regulation of protein processing
          innate immune response
          positive regulation of complement activation
          negative regulation of viral process
          positive regulation of phagocytosis
          defense response to Gram-positive bacterium
          protein complex oligomerization
          protein homooligomerization
          killing by host of symbiont cells
          protein homotrimerization
          protease binding
          signaling receptor binding
          calcium ion binding
          galactose binding
          mannose binding
          carbohydrate binding
          protein self-association
          calcium-dependent protein binding
          phosphatidylinositol-4-phosphate binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P104643 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 378233 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 294253 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 610890 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 644432 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 289244 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 247024 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 650703 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 588263 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 271514 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 159733 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 553400 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 113439 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 851891 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 362377 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 179345 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 9219 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P104643 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P104643 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P104643 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P401357 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 91799 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 61267 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10714786 - -
          Affymetrix Rat Gene 1.0 ST Array 10714783 - -
          Affymetrix Rat Gene 1.0 ST Array 10714787 - -
          Affymetrix Rat Gene 1.0 ST Array 10714784 - -
          Affymetrix Rat Gene 1.0 ST Array 10714785 - -
          Affymetrix Rat Gene 1.1 ST Array 10714782 - -
          Rat Genome 230 2.0 Array 1387528_PM_at - -
          Rat Genome 230 2.0 Array 1387528_at - -
          Affymetrix HT Rat Focus Array 1387528_at - -
          Affymetrix HT Rat Focus Array 1387528_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0100028177.rn.1 TC0100003465.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100028175.rn.1 TC0100003465.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100028173.rn.1 TC0100003465.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100028172.rn.1 TC0100003465.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100028174.rn.1 TC0100003465.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100028178.rn.1 TC0100003465.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100028176.rn.1 TC0100003465.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100028179.rn.1 TC0100003465.rn.1 RTA TC
          Rat Expression Set 230 1387528_PM_at - -
          Rat Expression Set 230 1387528_at - -
          Affymetrix Rat Gene 1.0 ST Array 10714782 - -
          Rat Genome U34 Set X05023_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21330470 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1374623 - -
          RatRef-12 Expression BeadChip ILMN_50869 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-97865cc7b-9fgnx:80/100.66.128.131:80.
          git-commit: d53bc399386730d6c5c6d01fe94b21e0ad3fdbfa
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0