Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00575617_m1
          See other CRHR2 GE Assays ›
          Gene Symbol:
          Crhr2
          Gene Name:
          corticotropin releasing hormone receptor 2
          Gene Aliases:
          Crf2r
          Chromosome Location:
          Chr.4: 85286371 - 85329374 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00575617_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Crhr2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_022714.1 NP_073205.1 5 - 6 755 82
          XM_006236556.2 XP_006236618.1 6 - 7 745 82
          XM_006236557.2 XP_006236619.1 6 - 7 742 82
          XM_006236558.1 XP_006236620.1 5 - 6 773 82
          GenBank mRNA EF078964.1 - 5 - 6 540 82
          U16253.1 - 5 - 6 755 82

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0400002950.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Crhr2

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          actin filament organization
          G-protein coupled receptor signaling pathway
          protein kinase C-activating G-protein coupled receptor signaling pathway
          feeding behavior
          hormone-mediated signaling pathway
          positive regulation of heart rate
          positive regulation of gene expression
          negative regulation of gene expression
          negative regulation of norepinephrine secretion
          positive regulation of serotonin secretion
          negative regulation of angiogenesis
          sensory perception of pain
          cerebellum development
          hypothalamus development
          positive regulation of cAMP metabolic process
          negative regulation of cAMP biosynthetic process
          positive regulation of cAMP biosynthetic process
          epithelial cell differentiation
          positive regulation of interleukin-6 production
          negative regulation of epinephrine secretion
          positive regulation of stress-activated MAPK cascade
          negative regulation of luteinizing hormone secretion
          gastric motility
          catecholamine biosynthetic process
          positive regulation of cAMP-mediated signaling
          negative regulation of cAMP-mediated signaling
          positive regulation of blood pressure
          negative regulation of follicle-stimulating hormone secretion
          skeletal muscle tissue growth
          positive regulation of NF-kappaB transcription factor activity
          long-term synaptic potentiation
          negative regulation of insulin secretion involved in cellular response to glucose stimulus
          regulation of ERK1 and ERK2 cascade
          cellular response to corticotropin-releasing hormone stimulus
          cellular response to glucocorticoid stimulus
          negative regulation of calcium ion import
          negative regulation of feeding behavior
          negative regulation of defecation
          G-protein coupled receptor activity
          hormone activity
          corticotrophin-releasing factor receptor activity
          peptide hormone binding
          corticotropin-releasing hormone receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P079538 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P079543 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 762961 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 97734 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 529940 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 911165 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 860336 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 807626 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 286520 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 495230 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 767119 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 325530 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 8867 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 614720 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 137375 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 32111 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 140855 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 72215 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 578878 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 747751 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 133311 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 160760 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 749936 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 714881 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 288387 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 844277 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 23068 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 10426 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P079538 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P079543 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P079543 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P079538 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P079543 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P079538 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P12312 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 3166 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 59803 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10862686 - -
          Affymetrix Rat Gene 1.0 ST Array 10862683 - -
          Affymetrix Rat Gene 1.0 ST Array 10862679 - -
          Affymetrix Rat Gene 1.0 ST Array 10862681 - -
          Affymetrix Rat Gene 1.0 ST Array 10862685 - -
          Affymetrix Rat Gene 1.0 ST Array 10862690 - -
          Affymetrix Rat Gene 1.0 ST Array 10862688 - -
          Affymetrix Rat Gene 1.0 ST Array 10862678 - -
          Affymetrix Rat Gene 1.0 ST Array 10862687 - -
          Affymetrix Rat Gene 1.0 ST Array 10862684 - -
          Affymetrix Rat Gene 1.0 ST Array 10862682 - -
          Affymetrix Rat Gene 1.0 ST Array 10862691 - -
          Affymetrix Rat Gene 1.0 ST Array 10862689 - -
          Affymetrix Rat Gene 1.0 ST Array 10862680 - -
          Affymetrix Rat Gene 1.1 ST Array 10862677 - -
          Rat Genome 230 2.0 Array 1369495_PM_at - -
          Rat Genome 230 2.0 Array 1369495_at - -
          Affymetrix HT Rat Focus Array 1369495_PM_at - -
          Affymetrix HT Rat Focus Array 1369495_at - -
          Affymetrix Rat Clariom S Assay PSR0400021012.rn.1 TC0400002950.rn.1 RTA TC
          Rat Expression Set 230 1369495_at - -
          Rat Expression Set 230 1369495_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10862677 - -
          Rat Genome U34 Set U16253_at - -
          Rat Genome U34 Set U16253_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20709808 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1357964 - -
          RatRef-12 Expression BeadChip ILMN_58814 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5777fb6d44-4pg6x:80/100.66.134.75:80.
          git-commit: 3a4c0ffdaddae765c0fdfab5c280296089abf14d
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0