Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00577043_m1
          See other SECISBP2 GE Assays ›
          Gene Symbol:
          Secisbp2
          Gene Name:
          SECIS binding protein 2
          Gene Aliases:
          Sbp2
          Chromosome Location:
          Chr.17: 13555348 - 13586601 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00577043_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Secisbp2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_024002.1 NP_076492.1 7 - 8 1105 75
          XM_008771506.1 XP_008769728.1 7 - 8 1155 75
          GenBank mRNA AJ251245.1 - 7 - 8 1105 75
          FQ231169.1 - 7 - 8 1127 75
          FQ232320.1 - 7 - 8 1133 75
          FQ234993.1 - 7 - 8 1149 75

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1700001397.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Secisbp2

          Gene Ontology Categories:

          Function(s) Process(es)

          selenocysteine incorporation
          striatum development
          neuron development
          negative regulation of nuclear-transcribed mRNA catabolic process, nonsense-mediated decay
          RNA binding
          mRNA 3'-UTR binding
          translation factor activity, RNA binding
          selenocysteine insertion sequence binding
          ribonucleoprotein complex binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P12396 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 575930 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 174258 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 308015 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 470030 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 791981 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 266842 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 573907 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 45423 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 334939 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 567590 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 498840 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 223344 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 156433 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 858985 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 505375 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 178987 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 229688 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 42294 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 421630 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 677976 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 751922 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 325896 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 308340 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 39523 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 71516 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 137174 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 53872 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 76625 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P12396 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P12396 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P12396 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P12396 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P252093 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701233725 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 28677 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 41702 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 99495 - -
          Agilent Rat cDNA Microarray (G4105A) 701233725 - -
          Agilent Rat cDNA Microarray (G4105A) 700300277 - -
          Agilent Rat cDNA Microarray (G4105A) 701714673 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10797555 - -
          Affymetrix Rat Gene 1.0 ST Array 10797542 - -
          Affymetrix Rat Gene 1.0 ST Array 10797547 - -
          Affymetrix Rat Gene 1.0 ST Array 10797543 - -
          Affymetrix Rat Gene 1.0 ST Array 10797546 - -
          Affymetrix Rat Gene 1.0 ST Array 10797549 - -
          Affymetrix Rat Gene 1.0 ST Array 10797550 - -
          Affymetrix Rat Gene 1.0 ST Array 10797551 - -
          Affymetrix Rat Gene 1.0 ST Array 10797558 - -
          Affymetrix Rat Gene 1.0 ST Array 10797557 - -
          Affymetrix Rat Gene 1.0 ST Array 10797554 - -
          Affymetrix Rat Gene 1.0 ST Array 10797556 - -
          Affymetrix Rat Gene 1.0 ST Array 10797544 - -
          Affymetrix Rat Gene 1.0 ST Array 10797552 - -
          Affymetrix Rat Gene 1.0 ST Array 10797553 - -
          Affymetrix Rat Gene 1.0 ST Array 10797545 - -
          Affymetrix Rat Gene 1.0 ST Array 10797548 - -
          Affymetrix Rat Gene 1.1 ST Array 10797541 - -
          Rat Genome 230 2.0 Array 1368094_at - -
          Rat Genome 230 2.0 Array 1368094_PM_at - -
          Affymetrix HT Rat Focus Array 1368094_at - -
          Affymetrix HT Rat Focus Array 1368094_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1700009749.rn.1 TC1700001397.rn.1 RTA TC
          Rat Expression Set 230 1368094_PM_at - -
          Rat Expression Set 230 1368094_at - -
          Affymetrix Rat Gene 1.0 ST Array 10797541 - -
          Rat Genome U34 Set rc_AI010977_at - -
          Rat Genome U34 Set rc_AI176759_at - -
          Rat Genome U34 Set rc_AA819474_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22087721 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1367121 - -
          RatRef-12 Expression BeadChip ILMN_58982 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0