Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00578590_m1
          See other SIGMAR1 GE Assays ›
          Gene Symbol:
          Sigmar1
          Gene Name:
          sigma non-opioid intracellular receptor 1
          Gene Aliases:
          Oprs1
          Chromosome Location:
          Chr.5: 58121826 - 58124604 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00578590_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Sigmar1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_030996.1 NP_112258.1 3 - 4 483 73
          GenBank mRNA AF004218.1 - 3 - 4 483 73
          AF067769.1 - 3 - 4 535 73
          AF087827.1 - 3 - 4 488 73
          BC061978.1 - 3 - 4 472 73

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0500002584.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Sigmar1

          Gene Ontology Categories:

          Function(s) Process(es)

          lipid transport
          nervous system development
          G protein-coupled opioid receptor signaling pathway
          regulation of neuron apoptotic process
          G protein-coupled opioid receptor activity
          protein binding
          signaling receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P609522 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P041323 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 213113 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 655140 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P041323 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P609522 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P609522 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P041323 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P041323 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P609522 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P609522 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P171634 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700505457 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 2182 - -
          Agilent Rat cDNA Microarray (G4105A) 700505457 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10876187 - -
          Affymetrix Rat Gene 1.0 ST Array 10876190 - -
          Affymetrix Rat Gene 1.0 ST Array 10876186 - -
          Affymetrix Rat Gene 1.0 ST Array 10876191 - -
          Affymetrix Rat Gene 1.0 ST Array 10876188 - -
          Affymetrix Rat Gene 1.0 ST Array 10876189 - -
          Affymetrix Rat Gene 1.1 ST Array 10876185 - -
          Rat Genome 230 2.0 Array 1386918_PM_a_at - -
          Rat Genome 230 2.0 Array 1386918_a_at - -
          Affymetrix HT Rat Focus Array 1386918_PM_a_at - -
          Affymetrix HT Rat Focus Array 1386918_a_at - -
          Affymetrix Rat Clariom S Assay PSR0500019076.rn.1 TC0500002584.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500019077.rn.1 TC0500002584.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500019075.rn.1 TC0500002584.rn.1 RTA TC
          Rat Expression Set 230 1386918_PM_a_at - -
          Rat Expression Set 230 1386918_a_at - -
          Affymetrix Rat Gene 1.0 ST Array 10876185 - -
          Rat Genome U34 Set AF004218_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21267503 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1365626 - -
          RatRef-12 Expression BeadChip ILMN_55721 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0