Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00579848_m1
          See other MDH2 GE Assays ›
          Gene Symbol:
          Mdh2
          Gene Name:
          malate dehydrogenase 2, NAD (mitochondrial)
          Gene Aliases:
          Mor1
          Chromosome Location:
          Chr.12: 23941451 - 23954406 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00579848_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mdh2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_031151.2 NP_112413.2 5 - 6 655 86
          GenBank mRNA BC063165.1 - 5 - 6 655 86
          DQ402951.1 - 4 - 5 489 86
          FQ213494.1 - 5 - 6 608 86
          FQ213817.1 - 5 - 6 570 86
          FQ215385.1 - 5 - 6 615 86
          FQ215647.1 - 5 - 6 616 86
          FQ216782.1 - 5 - 6 646 86
          FQ217232.1 - 5 - 6 607 86
          FQ224847.1 - 5 - 6 552 86
          X04240.1 - 5 - 6 596 86

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1200001345.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Mdh2

          Gene Ontology Categories:

          Function(s) Process(es)

          tricarboxylic acid cycle
          oxaloacetate metabolic process
          malate metabolic process
          internal protein amino acid acetylation
          NADH metabolic process
          cellular carbohydrate metabolic process
          RNA binding
          protein binding
          malate dehydrogenase activity
          L-malate dehydrogenase activity
          protein self-association
          malate dehydrogenase (NADP+) activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P413701 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 104169 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 307792 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 421254 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 769519 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 407865 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 36806 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 89168 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 872764 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 488389 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 447344 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 893544 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 51848 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 128176 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 103977 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P413701 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P413701 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P413701 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P413701 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700629623 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 46903 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 52544 - -
          Agilent Rat cDNA Microarray (G4105A) 700629623 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10761156 - -
          Affymetrix Rat Gene 1.0 ST Array 10761158 - -
          Affymetrix Rat Gene 1.0 ST Array 10761153 - -
          Affymetrix Rat Gene 1.0 ST Array 10761154 - -
          Affymetrix Rat Gene 1.0 ST Array 10761161 - -
          Affymetrix Rat Gene 1.0 ST Array 10761155 - -
          Affymetrix Rat Gene 1.0 ST Array 10761159 - -
          Affymetrix Rat Gene 1.0 ST Array 10761160 - -
          Affymetrix Rat Gene 1.0 ST Array 10761157 - -
          Affymetrix Rat Gene 1.1 ST Array 10761152 - -
          Rat Genome 230 2.0 Array 1369927_PM_at - -
          Rat Genome 230 2.0 Array 1369927_at - -
          Affymetrix HT Rat Focus Array 1369927_at - -
          Affymetrix HT Rat Focus Array 1369927_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1200011561.rn.1 TC1200001345.rn.1 RTA TC
          Rat Expression Set 230 1369927_at - -
          Rat Expression Set 230 1369927_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10761152 - -
          Rat Genome U34 Set rc_AI010480_at - -
          Rat Genome U34 Set rc_AI010480_g_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22117715 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1374806 - -
          RatRef-12 Expression BeadChip ILMN_57535 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-8696d955b-mh7vc:80/100.66.135.11:80.
          git-commit: b6496d4449ebd8b46e03b29da6eff4841d0bfc13
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-Offline