Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00580060_m1
          See other BCL10 GE Assays ›
          Gene Symbol:
          Bcl10
          Gene Name:
          B-cell CLL/lymphoma 10
          Gene Aliases:
          -
          Chromosome Location:
          Chr.2: 251805392 - 251814957 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00580060_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Bcl10
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_031328.1 NP_112618.1 2 - 3 477 76
          GenBank mRNA AB016069.1 - 2 - 3 477 76
          BC061772.1 - 2 - 3 465 76

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0200002364.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Bcl10

          Gene Ontology Categories:

          Function(s) Process(es)

          B cell apoptotic process
          neural tube closure
          response to molecule of bacterial origin
          adaptive immune response
          negative regulation of mature B cell apoptotic process
          apoptotic process
          cellular defense response
          I-kappaB kinase/NF-kappaB signaling
          cell death
          response to fungus
          immunoglobulin mediated immune response
          positive regulation of protein ubiquitination
          response to food
          positive regulation of phosphorylation
          positive regulation of apoptotic process
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          positive regulation of cysteine-type endopeptidase activity involved in apoptotic process
          fibroblast apoptotic process
          innate immune response
          positive regulation of transcription, DNA-templated
          T cell receptor signaling pathway
          regulation of T cell receptor signaling pathway
          positive regulation of T cell activation
          positive regulation of NF-kappaB transcription factor activity
          protein complex oligomerization
          protein homooligomerization
          protein heterooligomerization
          T cell apoptotic process
          thymocyte apoptotic process
          cellular response to mechanical stimulus
          positive regulation of extrinsic apoptotic signaling pathway
          protease binding
          transcription coactivator activity
          protein binding
          protein C-terminus binding
          transcription factor binding
          kinase activator activity
          enzyme binding
          kinase binding
          protein kinase binding
          ubiquitin protein ligase binding
          ubiquitin binding
          protein kinase B binding
          protein self-association
          NF-kappaB binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P264102 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 23799 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 564297 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 228811 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 347203 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 311670 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 67148 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P264102 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P264102 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P264102 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P264102 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700311502 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 89531 - -
          Agilent Rat cDNA Microarray (G4105A) 700311502 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10819656 - -
          Affymetrix Rat Gene 1.0 ST Array 10819654 - -
          Affymetrix Rat Gene 1.0 ST Array 10819655 - -
          Affymetrix Rat Gene 1.0 ST Array 10819657 - -
          Affymetrix Rat Gene 1.1 ST Array 10819653 - -
          Rat Genome 230 2.0 Array 1368118_PM_at - -
          Rat Genome 230 2.0 Array 1368118_at - -
          Affymetrix HT Rat Focus Array 1368118_PM_at - -
          Affymetrix HT Rat Focus Array 1368118_at - -
          Affymetrix Rat Clariom S Assay PSR0200016555.rn.1 TC0200002364.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200016556.rn.1 TC0200002364.rn.1 RTA TC
          Rat Expression Set 230 1368118_at - -
          Rat Expression Set 230 1368118_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10819653 - -
          Rat Genome U34 Set rc_AI104659_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22124924 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1350965 - -
          RatRef-12 Expression BeadChip ILMN_64237 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-59df64f8c5-6gx82:80/100.66.130.62:80.
          git-commit: fe83b6adba5b7aa08a343fc2038c841d76c6a25b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0