Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00581351_m1
          See other RABGGTA GE Assays ›
          Gene Symbol:
          Rabggta
          Gene Name:
          Rab geranylgeranyltransferase, alpha subunit
          Gene Aliases:
          -
          Chromosome Location:
          Chr.15: 34394074 - 34400468 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00581351_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rabggta
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_031654.2 NP_113842.1 6 - 7 722 88
          XM_006252009.2 XP_006252071.1 6 - 7 817 88
          XM_006252010.2 XP_006252072.1 6 - 7 764 88
          GenBank mRNA BC086547.1 - 6 - 7 722 88
          L10415.1 - 682 88
          S62096.1 - 682 88

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1500001720.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rabggta

          Gene Ontology Categories:

          Function(s) Process(es)

          protein geranylgeranylation
          Rab geranylgeranyltransferase activity
          zinc ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P418694 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 779056 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 289853 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 503419 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 184812 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 783073 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 70815 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P418694 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P418694 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P418694 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P652202 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P418694 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700376634 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 43360 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 18883 - -
          Agilent Rat cDNA Microarray (G4105A) 700376634 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10783911 - -
          Affymetrix Rat Gene 1.0 ST Array 10783901 - -
          Affymetrix Rat Gene 1.0 ST Array 10783914 - -
          Affymetrix Rat Gene 1.0 ST Array 10783905 - -
          Affymetrix Rat Gene 1.0 ST Array 10783906 - -
          Affymetrix Rat Gene 1.0 ST Array 10783907 - -
          Affymetrix Rat Gene 1.0 ST Array 10783916 - -
          Affymetrix Rat Gene 1.0 ST Array 10783915 - -
          Affymetrix Rat Gene 1.0 ST Array 10783912 - -
          Affymetrix Rat Gene 1.0 ST Array 10783903 - -
          Affymetrix Rat Gene 1.0 ST Array 10783902 - -
          Affymetrix Rat Gene 1.0 ST Array 10783899 - -
          Affymetrix Rat Gene 1.0 ST Array 10783910 - -
          Affymetrix Rat Gene 1.0 ST Array 10783904 - -
          Affymetrix Rat Gene 1.0 ST Array 10783913 - -
          Affymetrix Rat Gene 1.0 ST Array 10783900 - -
          Affymetrix Rat Gene 1.0 ST Array 10783909 - -
          Affymetrix Rat Gene 1.1 ST Array 10783898 - -
          Rat Genome 230 2.0 Array 1387869_s_at - -
          Rat Genome 230 2.0 Array 1395727_at - -
          Rat Genome 230 2.0 Array 1369899_PM_s_at - -
          Rat Genome 230 2.0 Array 1395727_PM_at - -
          Rat Genome 230 2.0 Array 1369899_s_at - -
          Rat Genome 230 2.0 Array 1387869_PM_s_at - -
          Affymetrix HT Rat Focus Array 1369899_PM_s_at - -
          Affymetrix HT Rat Focus Array 1395727_PM_at - -
          Affymetrix HT Rat Focus Array 1387869_PM_s_at - -
          Affymetrix HT Rat Focus Array 1387869_s_at - -
          Affymetrix HT Rat Focus Array 1369899_s_at - -
          Affymetrix HT Rat Focus Array 1395727_at - -
          Affymetrix Rat Clariom S Assay PSR1500011224.rn.1 TC1500001720.rn.1 RTA TC
          Rat Expression Set 230 1369899_PM_s_at - -
          Rat Expression Set 230 1387869_s_at - -
          Rat Expression Set 230 1387869_PM_s_at - -
          Rat Expression Set 230 1369899_s_at - -
          Rat Expression Set 230 1395727_at - -
          Rat Expression Set 230 1395727_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10783898 - -
          Rat Genome U34 Set S62096_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21723885 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1349529 - -
          RatRef-12 Expression BeadChip ILMN_62101 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0