Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00581565_m1
          See other FHL2 GE Assays ›
          Gene Symbol:
          Fhl2
          Gene Name:
          four and a half LIM domains 2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.9: 49920611 - 49964902 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00581565_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Fhl2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_031677.1 NP_113865.1 2 - 3 327 64
          XM_006244849.2 XP_006244911.1 3 - 4 2464 64
          GenBank mRNA AB008571.1 - 2 - 3 327 64

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0900001907.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Fhl2

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription by RNA polymerase II
          osteoblast differentiation
          transcription, DNA-templated
          regulation of transcription by RNA polymerase II
          response to hormone
          negative regulation of apoptotic process
          negative regulation of transcription, DNA-templated
          atrial cardiac muscle cell development
          ventricular cardiac muscle cell development
          heart trabecula formation
          protein binding
          transcription factor binding
          zinc ion binding
          identical protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P390700 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 517324 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 247492 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 871332 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 668669 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 707302 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 681204 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 139442 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 171389 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 936367 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 438532 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 477600 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 468994 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 679941 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 153187 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 314420 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 440096 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 161754 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 831333 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 934818 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 567827 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 67944 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 741896 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 101863 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 916 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 60629 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 16975 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 105286 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P390700 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P390700 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P390700 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P390700 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701623405 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 22833 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 68193 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 82950 - -
          Agilent Rat cDNA Microarray (G4105A) 701623405 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10927694 - -
          Affymetrix Rat Gene 1.0 ST Array 10927696 - -
          Affymetrix Rat Gene 1.0 ST Array 10927693 - -
          Affymetrix Rat Gene 1.0 ST Array 10927697 - -
          Affymetrix Rat Gene 1.0 ST Array 10927695 - -
          Affymetrix Rat Gene 1.1 ST Array 10927692 - -
          Rat Genome 230 2.0 Array 1369313_PM_at - -
          Rat Genome 230 2.0 Array 1369313_at - -
          Rat Genome 230 2.0 Array 1371951_at - -
          Rat Genome 230 2.0 Array 1371951_PM_at - -
          Affymetrix HT Rat Focus Array 1369313_at - -
          Affymetrix HT Rat Focus Array 1371951_PM_at - -
          Affymetrix HT Rat Focus Array 1369313_PM_at - -
          Affymetrix HT Rat Focus Array 1371951_at - -
          Affymetrix Rat Clariom S Assay PSR0900015646.rn.1 TC0900001907.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900015649.rn.1 TC0900001907.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900015647.rn.1 TC0900001907.rn.1 RTA TC
          Rat Expression Set 230 1371951_PM_at - -
          Rat Expression Set 230 1369313_PM_at - -
          Rat Expression Set 230 1371951_at - -
          Rat Expression Set 230 1369313_at - -
          Affymetrix Rat Gene 1.0 ST Array 10927692 - -
          Rat Genome U34 Set rc_AA891527_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21480977 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1372988 - -
          RatRef-12 Expression BeadChip ILMN_70315 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0