Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00581647_m1
          See other SCN7A GE Assays ›
          Gene Symbol:
          Scn7a
          Gene Name:
          sodium channel, voltage-gated, type VII, alpha subunit
          Gene Aliases:
          Na-G, Scn6a
          Chromosome Location:
          Chr.3: 52775779 - 52877319 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00581647_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Scn7a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_031686.1 NP_113874.1 7 - 8 950 72
          XM_006234296.2 XP_006234358.1 8 - 9 1227 72
          XM_006234298.2 XP_006234360.1 8 - 9 1227 72
          GenBank mRNA Y09164.1 - 7 - 8 950 72

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300003233.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Scn7a

          Gene Ontology Categories:

          Function(s) Process(es)

          sodium ion transport
          muscle contraction
          neuronal action potential
          regulation of ion transmembrane transport
          sodium ion transmembrane transport
          sodium ion homeostasis
          membrane depolarization during action potential
          voltage-gated sodium channel activity
          sodium channel activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P804822 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P026716 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 927808 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 337155 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 725880 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 868811 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 419505 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 468473 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 482381 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 572311 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 719585 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 409390 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 283429 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 284120 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 499695 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 133779 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 756246 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 30132 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 25078 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 506855 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 112286 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 205989 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 165180 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 368626 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 729770 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 780131 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 604274 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 906077 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 822950 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 222850 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 96207 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 340307 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 476843 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 229565 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 77796 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 645545 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 20129 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 954956 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 272159 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 165790 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 960692 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 808219 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 659841 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 375436 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 232037 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 435654 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 530702 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 616480 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 876024 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 346636 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 701730 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 135257 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 101224 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 179304 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 106691 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 78789 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 46635 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 115466 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 85327 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P804822 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P026716 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P804822 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P026716 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P026716 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P804822 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P804822 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700416341 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 9360 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 55221 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 19400 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 68833 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 51443 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 18650 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 90205 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 58351 - -
          Agilent Rat cDNA Microarray (G4105A) 700416341 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10845923 - -
          Affymetrix Rat Gene 1.0 ST Array 10845930 - -
          Affymetrix Rat Gene 1.0 ST Array 10845921 - -
          Affymetrix Rat Gene 1.0 ST Array 10845929 - -
          Affymetrix Rat Gene 1.0 ST Array 10845943 - -
          Affymetrix Rat Gene 1.0 ST Array 10845936 - -
          Affymetrix Rat Gene 1.0 ST Array 10845931 - -
          Affymetrix Rat Gene 1.0 ST Array 10845938 - -
          Affymetrix Rat Gene 1.0 ST Array 10845920 - -
          Affymetrix Rat Gene 1.0 ST Array 10845919 - -
          Affymetrix Rat Gene 1.0 ST Array 10845927 - -
          Affymetrix Rat Gene 1.0 ST Array 10845934 - -
          Affymetrix Rat Gene 1.0 ST Array 10845933 - -
          Affymetrix Rat Gene 1.0 ST Array 10845924 - -
          Affymetrix Rat Gene 1.0 ST Array 10845928 - -
          Affymetrix Rat Gene 1.0 ST Array 10845941 - -
          Affymetrix Rat Gene 1.0 ST Array 10845944 - -
          Affymetrix Rat Gene 1.0 ST Array 10845925 - -
          Affymetrix Rat Gene 1.0 ST Array 10845918 - -
          Affymetrix Rat Gene 1.0 ST Array 10845932 - -
          Affymetrix Rat Gene 1.0 ST Array 10845922 - -
          Affymetrix Rat Gene 1.0 ST Array 10845935 - -
          Affymetrix Rat Gene 1.0 ST Array 10845939 - -
          Affymetrix Rat Gene 1.0 ST Array 10845937 - -
          Affymetrix Rat Gene 1.0 ST Array 10845926 - -
          Affymetrix Rat Gene 1.0 ST Array 10845940 - -
          Affymetrix Rat Gene 1.1 ST Array 10845917 - -
          Rat Genome 230 2.0 Array 1370973_at - -
          Rat Genome 230 2.0 Array 1370973_PM_at - -
          Affymetrix HT Rat Focus Array 1370973_at - -
          Affymetrix HT Rat Focus Array 1370973_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0300025163.rn.1 TC0300003233.rn.1 RTA TC
          Rat Expression Set 230 1370973_at - -
          Rat Expression Set 230 1370973_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10845917 - -
          Rat Genome U34 Set rc_AA925248_at - -
          Rat Genome U34 Set S75991_at - -
          Rat Genome U34 Set rc_AA925248_at - -
          Rat Genome U34 Set S75991_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21868382 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1351576 - -
          RatRef-12 Expression BeadChip ILMN_53535 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5777fb6d44-4pg6x:80/100.66.134.75:80.
          git-commit: 3a4c0ffdaddae765c0fdfab5c280296089abf14d
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0