Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00582193_m1
          See other FIGF GE Assays ›
          Gene Symbol:
          Figf
          Gene Name:
          c-fos induced growth factor
          Gene Aliases:
          Vegf-d
          Chromosome Location:
          Chr.X: 31816482 - 31852278 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00582193_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Figf
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_031761.1 NP_113949.1 5 - 6 1025 91
          XM_006256863.2 XP_006256925.1 6 - 7 1172 91
          XM_006256864.2 XP_006256926.1 5 - 6 1502 91
          GenBank mRNA AF014827.1 - 5 - 6 1025 91
          AY032728.1 - 5 - 6 800 91

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0X00001634.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Recommended Assay Rattus norvegicus Rat Tumor Metastasis Panel 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Figf

          Gene Ontology Categories:

          Function(s) Process(es)

          angiogenesis
          response to hypoxia
          cell population proliferation
          positive regulation of cell population proliferation
          cell differentiation
          regulation of vascular endothelial growth factor receptor signaling pathway
          positive regulation of interleukin-6 production
          positive regulation of angiogenesis
          vascular endothelial growth factor receptor signaling pathway
          positive chemotaxis
          induction of positive chemotaxis
          positive regulation of cell division
          positive regulation of mast cell chemotaxis
          response to interleukin-1
          vascular endothelial growth factor receptor binding
          growth factor activity
          chemoattractant activity
          protein homodimerization activity
          vascular endothelial growth factor receptor 2 binding
          vascular endothelial growth factor receptor 3 binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P469554 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 99935 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 450853 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 261420 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 140403 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 885184 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 379214 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 243906 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 967681 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 116247 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 33555 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 4381 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 193559 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 705492 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 640201 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 882379 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 667186 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 230405 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 953895 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 610992 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 37672 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 278422 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 268913 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 344927 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 121603 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 28744 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 74200 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 99132 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P469554 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P469554 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P469554 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P469554 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 56699 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 96806 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 70305 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 96782 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10937730 - -
          Affymetrix Rat Gene 1.0 ST Array 10937733 - -
          Affymetrix Rat Gene 1.0 ST Array 10937732 - -
          Affymetrix Rat Gene 1.0 ST Array 10937729 - -
          Affymetrix Rat Gene 1.0 ST Array 10937731 - -
          Affymetrix Rat Gene 1.0 ST Array 10937728 - -
          Affymetrix Rat Gene 1.0 ST Array 10937726 - -
          Affymetrix Rat Gene 1.0 ST Array 10937727 - -
          Affymetrix Rat Gene 1.1 ST Array 10937725 - -
          Rat Genome 230 2.0 Array 1387709_at - -
          Rat Genome 230 2.0 Array 1373882_PM_at - -
          Rat Genome 230 2.0 Array 1387709_PM_at - -
          Rat Genome 230 2.0 Array 1373882_at - -
          Affymetrix HT Rat Focus Array 1387709_at - -
          Affymetrix HT Rat Focus Array 1373882_PM_at - -
          Affymetrix HT Rat Focus Array 1373882_at - -
          Affymetrix HT Rat Focus Array 1387709_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0X00010588.rn.1 TC0X00001634.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0X00010587.rn.1 TC0X00001634.rn.1 RTA TC
          Rat Expression Set 230 1373882_PM_at - -
          Rat Expression Set 230 1387709_PM_at - -
          Rat Expression Set 230 1373882_at - -
          Rat Expression Set 230 1387709_at - -
          Affymetrix Rat Gene 1.0 ST Array 10937725 - -
          Rat Genome U34 Set AF014827_at - -
          Rat Genome U34 Set AA848639_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21858227 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1363525 - -
          RatRef-12 Expression BeadChip ILMN_50127 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0