Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00584162_g1
          See other TXN2 GE Assays ›
          Gene Symbol:
          Txn2
          Gene Name:
          thioredoxin 2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.7: 119144350 - 119158173 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00584162_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Txn2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053331.2 NP_445783.1 1 - 2 68 84
          GenBank mRNA BC081760.1 - 1 - 2 68 84
          FQ214521.1 - 1 - 2 94 84
          FQ214678.1 - 1 - 2 94 84
          FQ214699.1 - 1 - 2 74 84
          FQ214706.1 - 1 - 2 106 84
          FQ214836.1 - 1 - 2 94 84
          FQ215775.1 - 1 - 2 103 84
          FQ216090.1 - 1 - 2 74 84
          FQ216404.1 - 1 - 2 72 84
          FQ217268.1 - 1 - 2 72 84
          FQ220081.1 - 1 - 2 106 84
          FQ223556.1 - 1 - 2 81 84
          U73525.1 - 1 - 2 62 84

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0700003442.rn.1

          Additional Information:

          This assay is in the 5'UTR region of the BC081760.1, NM_053331.2, U73525.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Txn2

          Gene Ontology Categories:

          Function(s) Process(es)

          response to hypoxia
          glycerol ether metabolic process
          response to oxidative stress
          response to nutrient
          response to xenobiotic stimulus
          response to hormone
          response to glucose
          response to organic cyclic compound
          cellular response to nutrient levels
          cell redox homeostasis
          response to axon injury
          protein binding
          peptide-methionine (S)-S-oxide reductase activity
          protein-disulfide reductase activity
          peptide-methionine (R)-S-oxide reductase activity
          protein-containing complex binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P130641 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 813359 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 37173 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 605707 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 766862 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 583064 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 567425 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 957915 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 2846 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 337740 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 22246 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 20007 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 166536 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 120910 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P130641 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P130641 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P130641 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P130641 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700183127 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 90206 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 43947 - -
          Agilent Rat cDNA Microarray (G4105A) 700183127 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10905237 - -
          Affymetrix Rat Gene 1.0 ST Array 10905238 - -
          Affymetrix Rat Gene 1.0 ST Array 10905236 - -
          Affymetrix Rat Gene 1.0 ST Array 10905235 - -
          Affymetrix Rat Gene 1.0 ST Array 10905234 - -
          Affymetrix Rat Gene 1.1 ST Array 10905233 - -
          Rat Genome 230 2.0 Array 1398844_at - -
          Rat Genome 230 2.0 Array 1398844_PM_at - -
          Affymetrix HT Rat Focus Array 1398844_PM_at - -
          Affymetrix HT Rat Focus Array 1398844_at - -
          Affymetrix Rat Clariom S Assay PSR0700027015.rn.1 TC0700003442.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700027019.rn.1 TC0700003442.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700027013.rn.1 TC0700003442.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700027014.rn.1 TC0700003442.rn.1 RTA TC
          Rat Expression Set 230 1398844_PM_at - -
          Rat Expression Set 230 1398844_at - -
          Affymetrix Rat Gene 1.0 ST Array 10905233 - -
          Rat Genome U34 Set U73525_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21692068 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1368589 - -
          RatRef-12 Expression BeadChip ILMN_63603 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0