Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00584523_m1
          See other ATP1B4 GE Assays ›
          Gene Symbol:
          Atp1b4
          Gene Name:
          ATPase, (Na+)/K+ transporting, beta 4 polypeptide
          Gene Aliases:
          Ac2-628
          Chromosome Location:
          Chr.X: 124631543 - 124652520 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00584523_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Atp1b4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053381.1 NP_445833.1 6 - 7 891 84
          XM_006257493.2 XP_006257555.1 6 - 7 1182 84
          XM_006257494.2 XP_006257556.1 6 - 7 2616 84
          XM_006257495.2 XP_006257557.1 6 - 7 2601 84
          GenBank mRNA AF158385.2 - 6 - 7 891 84
          AF158386.2 - 6 - 7 879 84

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0X00001082.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Atp1b4

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          potassium ion transport
          sodium ion transport
          protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P12790 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 413975 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 543389 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 158809 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 111061 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 249519 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 913278 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 324887 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 624985 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 192752 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 786350 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 135881 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 465173 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 280060 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 827910 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 696269 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 800991 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 261463 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 130177 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 437218 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 790592 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 569018 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 808193 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 21067 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 60097 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 681646 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 157859 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 52262 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 123048 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 176331 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P12790 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P12790 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P12790 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P494089 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P12790 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 49596 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 29191 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 45106 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 10307 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 13475 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 18262 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10936253 - -
          Affymetrix Rat Gene 1.0 ST Array 10936252 - -
          Affymetrix Rat Gene 1.0 ST Array 10936254 - -
          Affymetrix Rat Gene 1.0 ST Array 10936257 - -
          Affymetrix Rat Gene 1.0 ST Array 10936256 - -
          Affymetrix Rat Gene 1.0 ST Array 10936251 - -
          Affymetrix Rat Gene 1.0 ST Array 10936258 - -
          Affymetrix Rat Gene 1.0 ST Array 10936249 - -
          Affymetrix Rat Gene 1.0 ST Array 10936250 - -
          Affymetrix Rat Gene 1.1 ST Array 10936248 - -
          Rat Genome 230 2.0 Array 1387321_PM_at - -
          Rat Genome 230 2.0 Array 1387321_at - -
          Affymetrix HT Rat Focus Array 1387321_PM_at - -
          Affymetrix HT Rat Focus Array 1387321_at - -
          Affymetrix Rat Clariom S Assay PSR0X00006669.rn.1 TC0X00001082.rn.1 RTA TC
          Rat Expression Set 230 1387321_at - -
          Rat Expression Set 230 1387321_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10936248 - -
          Rat Genome U34 Set rc_AI071721_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21657831 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1364076 - -
          RatRef-12 Expression BeadChip ILMN_59069 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0