Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00585220_m1
          See other CNGA1 GE Assays ›
          Gene Symbol:
          Cnga1
          Gene Name:
          cyclic nucleotide gated channel alpha 1
          Gene Aliases:
          Cncg, HCN
          Chromosome Location:
          Chr.14: 38155771 - 38171107 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00585220_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cnga1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053497.1 NP_445949.1 6 - 7 527 133
          GenBank mRNA U93851.1 - 6 - 7 527 133

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1400000450.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cnga1

          Gene Ontology Categories:

          Function(s) Process(es)

          spermatogenesis
          visual perception
          phototransduction, visible light
          ion transmembrane transport
          regulation of cytosolic calcium ion concentration
          membrane depolarization
          potassium ion transmembrane transport
          ion channel activity
          intracellular cyclic nucleotide activated cation channel activity
          intracellular cAMP-activated cation channel activity
          intracellular cGMP-activated cation channel activity
          voltage-gated potassium channel activity
          cGMP binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P030075 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 506207 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 804317 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 717007 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 321647 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 492354 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 122954 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 115996 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 71417 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 798943 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 888258 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 660070 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 139358 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 40618 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 134962 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P030075 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P030075 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P030075 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P12824 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 64913 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 71440 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10772452 - -
          Affymetrix Rat Gene 1.0 ST Array 10772449 - -
          Affymetrix Rat Gene 1.0 ST Array 10772454 - -
          Affymetrix Rat Gene 1.0 ST Array 10772456 - -
          Affymetrix Rat Gene 1.0 ST Array 10772450 - -
          Affymetrix Rat Gene 1.0 ST Array 10772455 - -
          Affymetrix Rat Gene 1.0 ST Array 10772453 - -
          Affymetrix Rat Gene 1.0 ST Array 10772451 - -
          Affymetrix Rat Gene 1.1 ST Array 10772448 - -
          Rat Genome 230 2.0 Array 1370013_PM_at - -
          Rat Genome 230 2.0 Array 1370013_at - -
          Affymetrix HT Rat Focus Array 1370013_PM_at - -
          Affymetrix HT Rat Focus Array 1370013_at - -
          Affymetrix Rat Clariom S Assay PSR1400003578.rn.1 TC1400000450.rn.1 RTA TC
          Rat Expression Set 230 1370013_PM_at - -
          Rat Expression Set 230 1370013_at - -
          Affymetrix Rat Gene 1.0 ST Array 10772448 - -
          Rat Genome U34 Set U76220_at - -
          Rat Genome U34 Set L02634_s_at - -
          Rat Genome U34 Set L02634_s_at - -
          Rat Genome U34 Set U76220_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21958039 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1352813 - -
          RatRef-12 Expression BeadChip ILMN_58989 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-845544bffd-zxsgg:80/100.66.135.11:80.
          git-commit: 2bb44be0f35b85de1c1d8e4f377b85e7a3a71488
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0