Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00589211_m1
          See other HNRNPU GE Assays ›
          Gene Symbol:
          Hnrnpu
          Gene Name:
          heterogeneous nuclear ribonucleoprotein U
          Gene Aliases:
          Hnrpu, SN1
          Chromosome Location:
          Chr.13: 96222095 - 96238850 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00589211_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Hnrnpu
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_057139.2 NP_476480.2 1 - 2 784 113
          XM_006250311.2 XP_006250373.1 1 - 2 1160 113
          GenBank mRNA BC072529.1 - 1 - 2 784 113
          D14048.1 - 1 - 2 1263 113

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1300002033.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Hnrnpu

          Gene Ontology Categories:

          Function(s) Process(es)

          osteoblast differentiation
          transport
          circadian regulation of gene expression
          CRD-mediated mRNA stabilization
          core promoter sequence-specific DNA binding
          RNA binding
          transporter activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P496211 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 75514 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 652798 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 75895 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 811499 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 251054 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 543737 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 114165 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P496211 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P496211 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P496211 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P254413 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P496211 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P496216 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 39113 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10770237 - -
          Affymetrix Rat Gene 1.0 ST Array 10770233 - -
          Affymetrix Rat Gene 1.0 ST Array 10770240 - -
          Affymetrix Rat Gene 1.0 ST Array 10770225 - -
          Affymetrix Rat Gene 1.0 ST Array 10770230 - -
          Affymetrix Rat Gene 1.0 ST Array 10770236 - -
          Affymetrix Rat Gene 1.0 ST Array 10770231 - -
          Affymetrix Rat Gene 1.0 ST Array 10770228 - -
          Affymetrix Rat Gene 1.0 ST Array 10770226 - -
          Affymetrix Rat Gene 1.0 ST Array 10770239 - -
          Affymetrix Rat Gene 1.0 ST Array 10770227 - -
          Affymetrix Rat Gene 1.0 ST Array 10770235 - -
          Affymetrix Rat Gene 1.0 ST Array 10770232 - -
          Affymetrix Rat Gene 1.0 ST Array 10770234 - -
          Affymetrix Rat Gene 1.0 ST Array 10770238 - -
          Affymetrix Rat Gene 1.0 ST Array 10770229 - -
          Affymetrix Rat Gene 1.1 ST Array 10770224 - -
          Rat Genome 230 2.0 Array 1370171_at - -
          Rat Genome 230 2.0 Array 1370170_PM_at - -
          Rat Genome 230 2.0 Array 1370171_PM_at - -
          Rat Genome 230 2.0 Array 1370170_at - -
          Affymetrix HT Rat Focus Array 1370171_at - -
          Affymetrix HT Rat Focus Array 1370170_at - -
          Affymetrix HT Rat Focus Array 1370171_PM_at - -
          Affymetrix HT Rat Focus Array 1370170_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1300014486.rn.1 TC1300002033.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300014487.rn.1 TC1300002033.rn.1 RTA TC
          Rat Expression Set 230 1370171_PM_at - -
          Rat Expression Set 230 1370170_PM_at - -
          Rat Expression Set 230 1370171_at - -
          Rat Expression Set 230 1370170_at - -
          Affymetrix Rat Gene 1.0 ST Array 10770224 - -
          Rat Genome U34 Set D14048_g_at - -
          Rat Genome U34 Set D14048_at - -
          Rat Genome U34 Set rc_AI058964_at - -
          Rat Genome U34 Set H32350_s_at - -
          Rat Genome U34 Set D14048_g_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20717614 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376257 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0