Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00589285_m1
          See other SEPT2 GE Assays ›
          Gene Symbol:
          Sept2
          Gene Name:
          septin 2
          Gene Aliases:
          Nedd5, Vesp11
          Chromosome Location:
          Chr.9: 100624848 - 100658053 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00589285_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Sept2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_057148.2 NP_476489.1 10 - 11 989 92
          XM_006245511.2 XP_006245573.1 10 - 11 1269 92
          XM_006245512.2 XP_006245574.1 10 - 11 1272 92
          XM_006245513.2 XP_006245575.1 10 - 11 1013 92
          XM_006245514.1 XP_006245576.1 10 - 11 1047 92
          GenBank mRNA AB027561.1 - 9 - 10 950 92
          BC081745.1 - 10 - 11 989 92
          FQ212097.1 - 10 - 11 1002 92

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0900001177.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Sept2

          Gene Ontology Categories:

          Function(s) Process(es)

          mitotic cell cycle
          regulation of L-glutamate import across plasma membrane
          smoothened signaling pathway
          metabolic process
          neuron projection development
          regulation of protein localization
          regulation of catalytic activity
          cell division
          cilium assembly
          GTPase activity
          GTP binding
          enzyme regulator activity
          molecular adaptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P147069 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 862658 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 356945 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 373288 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 241047 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 625805 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 59788 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 952005 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 40826 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 412890 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 738332 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 23767 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 604102 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 871683 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 834951 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 408162 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 664584 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 51402 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 971820 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 127857 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 250829 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 799787 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 883543 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 850152 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 55243 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 34921 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 111526 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 70783 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P147069 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P147069 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P147069 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P13047 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 79265 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 9402 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 23503 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10925608 - -
          Affymetrix Rat Gene 1.0 ST Array 10925607 - -
          Affymetrix Rat Gene 1.0 ST Array 10925601 - -
          Affymetrix Rat Gene 1.0 ST Array 10925605 - -
          Affymetrix Rat Gene 1.0 ST Array 10930048 - -
          Affymetrix Rat Gene 1.0 ST Array 10925606 - -
          Affymetrix Rat Gene 1.0 ST Array 10925603 - -
          Affymetrix Rat Gene 1.0 ST Array 10925604 - -
          Affymetrix Rat Gene 1.0 ST Array 10925602 - -
          Affymetrix Rat Gene 1.1 ST Array 10925600 - -
          Affymetrix Rat Gene 1.1 ST Array 10930047 - -
          Rat Genome 230 2.0 Array 1368984_at - -
          Rat Genome 230 2.0 Array 1368984_PM_at - -
          Rat Genome 230 2.0 Array 1388389_PM_at - -
          Rat Genome 230 2.0 Array 1388389_at - -
          Affymetrix HT Rat Focus Array 1388389_PM_at - -
          Affymetrix HT Rat Focus Array 1368984_PM_at - -
          Affymetrix HT Rat Focus Array 1368984_at - -
          Affymetrix HT Rat Focus Array 1388389_at - -
          Affymetrix Rat Clariom S Assay PSR0900010254.rn.1 TC0900001177.rn.1 RTA TC
          Rat Expression Set 230 1388389_PM_at - -
          Rat Expression Set 230 1388389_at - -
          Rat Expression Set 230 1368984_at - -
          Rat Expression Set 230 1368984_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10925600 - -
          Rat Genome U34 Set rc_AI179570_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20795674 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7b745768ff-vzrjf:80/100.66.128.131:80.
          git-commit: 4b95038805a43672018eef8c57cc3f15d8b0c074
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: integration/2.33.0-meta-desc