Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00589584_m1
          See other PCYT1A GE Assays ›
          Gene Symbol:
          Pcyt1a
          Gene Name:
          phosphate cytidylyltransferase 1, choline, alpha
          Gene Aliases:
          CCT-alpha, CTP
          Chromosome Location:
          Chr.11: 71547865 - 71592035 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00589584_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pcyt1a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_078622.2 NP_511177.2 5 - 6 590 78
          XM_006248444.2 XP_006248506.1 5 - 6 743 78
          XM_008768694.1 XP_008766916.1 5 - 6 941 78
          GenBank mRNA BC085713.1 - 5 - 6 588 78
          L13245.1 - 4 - 5 487 78
          M36071.1 - 5 - 6 587 78
          U03490.1 - 5 - 6 552 78

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1100001752.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Pcyt1a

          Gene Ontology Categories:

          Function(s) Process(es)

          phosphatidylcholine biosynthetic process
          CDP-choline pathway
          choline-phosphate cytidylyltransferase activity
          lipid binding
          protein homodimerization activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P705255 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P477116 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 454656 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 889399 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 503847 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 677201 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 102026 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 100213 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 319912 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 276489 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 899775 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 90999 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 203871 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 247470 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 886328 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 750502 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 84547 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 630546 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 231558 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 212089 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 214140 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 180555 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 647438 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 224126 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 885393 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 45532 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 769701 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 868891 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 158824 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 943621 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 6098 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 81919 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 105030 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 166745 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 44886 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 145148 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 89417 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P705255 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P477116 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P705255 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P477116 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P477116 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P705255 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P388310 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701490252 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 35519 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 24573 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 77899 - -
          Agilent Rat cDNA Microarray (G4105A) 701490252 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10754817 - -
          Affymetrix Rat Gene 1.0 ST Array 10754816 - -
          Affymetrix Rat Gene 1.0 ST Array 10754815 - -
          Affymetrix Rat Gene 1.0 ST Array 10754819 - -
          Affymetrix Rat Gene 1.0 ST Array 10754820 - -
          Affymetrix Rat Gene 1.0 ST Array 10754821 - -
          Affymetrix Rat Gene 1.0 ST Array 10754822 - -
          Affymetrix Rat Gene 1.0 ST Array 10754823 - -
          Affymetrix Rat Gene 1.0 ST Array 10754814 - -
          Affymetrix Rat Gene 1.0 ST Array 10754818 - -
          Affymetrix Rat Gene 1.1 ST Array 10754813 - -
          Rat Genome 230 2.0 Array 1369656_at - -
          Rat Genome 230 2.0 Array 1369656_PM_at - -
          Rat Genome 230 2.0 Array 1389299_at - -
          Rat Genome 230 2.0 Array 1389299_PM_at - -
          Affymetrix HT Rat Focus Array 1369656_PM_at - -
          Affymetrix HT Rat Focus Array 1389299_at - -
          Affymetrix HT Rat Focus Array 1389299_PM_at - -
          Affymetrix HT Rat Focus Array 1369656_at - -
          Affymetrix Rat Clariom S Assay PSR1100011067.rn.1 TC1100001752.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100011068.rn.1 TC1100001752.rn.1 RTA TC
          Rat Expression Set 230 1369656_at - -
          Rat Expression Set 230 1389299_at - -
          Rat Expression Set 230 1369656_PM_at - -
          Rat Expression Set 230 1389299_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10754813 - -
          Rat Genome U34 Set rc_AA925887_at - -
          Rat Genome U34 Set rc_AI011058_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22359080 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1370274 - -
          RatRef-12 Expression BeadChip ILMN_55341 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-75d45cbfdf-4zq6t:80/100.66.130.62:80.
          git-commit: a3c66eacce7c9fa3045440ddf4a86215532ee526
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix