Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00590124_m1
          See other FAIM GE Assays ›
          Gene Symbol:
          Faim LOC100362113
          Gene Name:
          Fas apoptotic inhibitory molecule
          Fas apoptotic inhibitory molecule
          Gene Aliases:
          -
          -
          Chromosome Location:
          Chr.8: 107223472 - 107241225 on Build Rnor_6.0
          Chromosome Location:
          Chr.8: 106809803 - 106827555 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Rn00590124_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Faim
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_080895.1 NP_543171.1 477 60
          XM_006243616.2 XP_006243678.1 624 60
          XM_006243617.2 XP_006243679.1 718 60
          XM_006243619.2 XP_006243681.1 661 60
          XM_006243620.2 XP_006243682.1 745 60
          XM_008766474.1 XP_008764696.1 834 60
          GenBank mRNA AF412334.1 - 477 60
          AF467453.1 - 458 60
          Gene Symbol: LOC100362113
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_002729959.3 XP_002730005.1 5 - 6 624 60
          XM_006243611.2 XP_006243673.1 5 - 6 718 60
          XM_006243613.2 XP_006243675.1 4 - 5 565 60
          XM_006243614.2 XP_006243676.1 4 - 5 661 60
          XM_006243615.2 XP_006243677.1 4 - 5 745 60
          XM_008766582.1 XP_008764804.1 5 - 6 834 60

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0800001360.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Faim
          Click here to find siRNAs for Gene LOC100362113

          Gene Ontology Categories:

          Function(s)

          apoptotic process
          I-kappaB kinase/NF-kappaB signaling
          negative regulation of apoptotic process
          positive regulation of neurogenesis
          negative regulation of extrinsic apoptotic signaling pathway via death domain receptors

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P604490 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P604490 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P604490 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P604490 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P604490 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10812335 - -
          Affymetrix Rat Gene 1.0 ST Array 10812337 - -
          Affymetrix Rat Gene 1.0 ST Array 10812333 - -
          Affymetrix Rat Gene 1.0 ST Array 10912462 - -
          Affymetrix Rat Gene 1.0 ST Array 10812334 - -
          Affymetrix Rat Gene 1.0 ST Array 10912458 - -
          Affymetrix Rat Gene 1.0 ST Array 10812332 - -
          Affymetrix Rat Gene 1.0 ST Array 10912460 - -
          Affymetrix Rat Gene 1.0 ST Array 10912463 - -
          Affymetrix Rat Gene 1.0 ST Array 10812338 - -
          Affymetrix Rat Gene 1.0 ST Array 10912457 - -
          Affymetrix Rat Gene 1.0 ST Array 10912459 - -
          Affymetrix Rat Gene 1.1 ST Array 10812331 - -
          Affymetrix Rat Gene 1.1 ST Array 10912456 - -
          Rat Genome 230 2.0 Array 1370044_at - -
          Rat Genome 230 2.0 Array 1370044_PM_at - -
          Affymetrix HT Rat Focus Array 1370044_PM_at - -
          Affymetrix HT Rat Focus Array 1370044_at - -
          Affymetrix Rat Clariom S Assay PSR0800009934.rn.1 TC0800001360.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800009933.rn.1 TC0800001360.rn.1 RTA TC
          Rat Expression Set 230 1370044_PM_at - -
          Rat Expression Set 230 1370044_at - -
          Affymetrix Rat Gene 1.0 ST Array 10912456 - -
          Affymetrix Rat Gene 1.0 ST Array 10812331 - -
          Rat Genome U34 Set rc_AI177251_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20830834 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_2040544 - -
          RatRef-12 Expression BeadChip ILMN_57271 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6467d75d44-xp7n2:80/100.66.130.62:80.
          git-commit: 8ea1bebb48d33b8b7c0e2103582aa482e6db8664
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0