Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00590996_m1
          See other FGF8 GE Assays ›
          Gene Symbol:
          Fgf8
          Gene Name:
          fibroblast growth factor 8
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 265492949 - 265498831 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00590996_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Fgf8
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_133286.1 NP_579820.1 3 - 4 219 66
          XM_006231436.2 XP_006231498.1 3 - 4 411 66
          XM_006231437.2 XP_006231499.1 4 - 5 339 66
          XM_008760398.1 XP_008758620.1 4 - 5 306 66
          GenBank mRNA AB079113.1 - 3 - 4 219 66

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100007798.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus Rat Androgen Panel 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Fgf8

          Gene Ontology Categories:

          Function(s) Process(es)

          MAPK cascade
          branching involved in blood vessel morphogenesis
          metanephros development
          branching involved in ureteric bud morphogenesis
          organ induction
          kidney development
          mesonephros development
          neural plate morphogenesis
          heart looping
          blood vessel remodeling
          heart morphogenesis
          outflow tract septum morphogenesis
          outflow tract morphogenesis
          epithelial to mesenchymal transition involved in endocardial cushion formation
          apoptotic process
          response to oxidative stress
          determination of left/right symmetry
          heart development
          motor neuron axon guidance
          mesodermal cell migration
          positive regulation of cell population proliferation
          gonad development
          fibroblast growth factor receptor signaling pathway
          response to xenobiotic stimulus
          embryo development ending in birth or egg hatching
          positive regulation of gene expression
          response to organic cyclic compound
          telencephalon development
          pallium development
          subpallium development
          forebrain dorsal/ventral pattern formation
          cell proliferation in forebrain
          forebrain neuron development
          central nervous system neuron development
          signal transduction involved in regulation of gene expression
          lung development
          BMP signaling pathway
          male genitalia development
          thyroid gland development
          otic vesicle formation
          midbrain-hindbrain boundary development
          dorsal/ventral axon guidance
          embryonic heart tube development
          limb morphogenesis
          embryonic hindlimb morphogenesis
          aorta morphogenesis
          inner ear morphogenesis
          odontogenesis
          regulation of odontogenesis of dentin-containing tooth
          negative regulation of apoptotic process
          negative regulation of neuron apoptotic process
          cell fate commitment
          positive regulation of mitotic nuclear division
          positive regulation of organ growth
          generation of neurons
          forebrain morphogenesis
          positive chemotaxis
          negative regulation of cardiac muscle tissue development
          pharyngeal system development
          canonical Wnt signaling pathway
          corticotropin hormone secreting cell differentiation
          thyroid-stimulating hormone-secreting cell differentiation
          bone development
          lung morphogenesis
          branching involved in salivary gland morphogenesis
          neuroepithelial cell differentiation
          dopaminergic neuron differentiation
          cell migration involved in mesendoderm migration
          type 1 fibroblast growth factor receptor binding
          type 2 fibroblast growth factor receptor binding
          growth factor activity
          chemoattractant activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P219639 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 302055 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 430005 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 277289 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 333695 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 130238 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P219639 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P219639 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P219639 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P219639 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10730438 - -
          Affymetrix Rat Gene 1.0 ST Array 10730439 - -
          Affymetrix Rat Gene 1.0 ST Array 10730440 - -
          Affymetrix Rat Gene 1.0 ST Array 10730445 - -
          Affymetrix Rat Gene 1.0 ST Array 10730444 - -
          Affymetrix Rat Gene 1.1 ST Array 10730437 - -
          Rat Genome 230 2.0 Array 1370117_at - -
          Rat Genome 230 2.0 Array 1370117_PM_at - -
          Affymetrix HT Rat Focus Array 1370117_PM_at - -
          Affymetrix HT Rat Focus Array 1370117_at - -
          Affymetrix Rat Clariom S Assay PSR0100062851.rn.1 TC0100007798.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100062852.rn.1 TC0100007798.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100062847.rn.1 TC0100007798.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100062846.rn.1 TC0100007798.rn.1 RTA TC
          Rat Expression Set 230 1370117_at - -
          Rat Expression Set 230 1370117_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10730437 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20876499 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1355059 - -
          RatRef-12 Expression BeadChip ILMN_48520 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0