Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00591325_m1
          See other LFNG GE Assays ›
          Gene Symbol:
          Lfng
          Gene Name:
          LFNG O-fucosylpeptide 3-beta-N-acetylglucosaminyltransferase
          Gene Aliases:
          -
          Chromosome Location:
          Chr.12: 16118192 - 16126211 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00591325_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Lfng
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_133393.2 NP_596884.1 1 - 2 488 63
          GenBank mRNA AB054539.1 - 1 - 2 429 63
          BC070933.1 - 1 - 2 488 63

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1200001174.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Lfng

          Gene Ontology Categories:

          Function(s) Process(es)

          ovarian follicle development
          somitogenesis
          female meiotic nuclear division
          compartment pattern specification
          biological_process
          metabolic process
          regulation of Notch signaling pathway
          regulation of somitogenesis
          positive regulation of protein binding
          positive regulation of Notch signaling pathway
          molecular_function
          O-fucosylpeptide 3-beta-N-acetylglucosaminyltransferase activity
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P152014 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 441719 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 656873 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 89468 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 276062 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 803951 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 378775 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 512707 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 133141 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P152014 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P152014 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P152014 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P13173 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 95147 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10760595 - -
          Affymetrix Rat Gene 1.0 ST Array 10760597 - -
          Affymetrix Rat Gene 1.0 ST Array 10760596 - -
          Affymetrix Rat Gene 1.0 ST Array 10760603 - -
          Affymetrix Rat Gene 1.0 ST Array 10760598 - -
          Affymetrix Rat Gene 1.0 ST Array 10760600 - -
          Affymetrix Rat Gene 1.0 ST Array 10760599 - -
          Affymetrix Rat Gene 1.0 ST Array 10760601 - -
          Affymetrix Rat Gene 1.0 ST Array 10760602 - -
          Affymetrix Rat Gene 1.0 ST Array 10760604 - -
          Affymetrix Rat Gene 1.1 ST Array 10760594 - -
          Rat Genome 230 2.0 Array 1389250_PM_at - -
          Rat Genome 230 2.0 Array 1370105_PM_at - -
          Rat Genome 230 2.0 Array 1370105_at - -
          Rat Genome 230 2.0 Array 1389250_at - -
          Affymetrix HT Rat Focus Array 1370105_at - -
          Affymetrix HT Rat Focus Array 1370105_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1200009968.rn.1 TC1200001174.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200009973.rn.1 TC1200001174.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200009974.rn.1 TC1200001174.rn.1 RTA TC
          Rat Expression Set 230 1370105_PM_at - -
          Rat Expression Set 230 1370105_at - -
          Affymetrix Rat Gene 1.0 ST Array 10760594 - -
          Rat Genome U34 Set rc_AI236775_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21279916 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1364471 - -
          RatRef-12 Expression BeadChip ILMN_57275 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0