Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00591594_m1
          See other ACTN3 GE Assays ›
          Gene Symbol:
          Actn3
          Gene Name:
          actinin alpha 3
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 220120532 - 220136460 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00591594_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Actn3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_133424.2 NP_596915.1 11 - 12 1311 88
          GenBank mRNA AF450248.1 - 11 - 12 1312 88
          AY096238.1 - 11 - 12 1236 88

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100007090.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Actn3

          Gene Ontology Categories:

          Function(s) Process(es)

          muscle contraction
          focal adhesion assembly
          calcium ion binding
          actin filament binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P656495 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 250172 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 876627 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 744258 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 924050 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 854662 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 282818 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 50189 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 854004 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 506266 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 378772 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 527593 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 957664 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 386819 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 170594 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 84054 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P656495 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P656495 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P656495 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P656495 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 9873 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 24990 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10727677 - -
          Affymetrix Rat Gene 1.0 ST Array 10727680 - -
          Affymetrix Rat Gene 1.0 ST Array 10727673 - -
          Affymetrix Rat Gene 1.0 ST Array 10727671 - -
          Affymetrix Rat Gene 1.0 ST Array 10727678 - -
          Affymetrix Rat Gene 1.0 ST Array 10727679 - -
          Affymetrix Rat Gene 1.0 ST Array 10727668 - -
          Affymetrix Rat Gene 1.0 ST Array 10727663 - -
          Affymetrix Rat Gene 1.0 ST Array 10727661 - -
          Affymetrix Rat Gene 1.0 ST Array 10727669 - -
          Affymetrix Rat Gene 1.0 ST Array 10727675 - -
          Affymetrix Rat Gene 1.0 ST Array 10727662 - -
          Affymetrix Rat Gene 1.0 ST Array 10727665 - -
          Affymetrix Rat Gene 1.0 ST Array 10727666 - -
          Affymetrix Rat Gene 1.0 ST Array 10727670 - -
          Affymetrix Rat Gene 1.0 ST Array 10727667 - -
          Affymetrix Rat Gene 1.0 ST Array 10727682 - -
          Affymetrix Rat Gene 1.0 ST Array 10727676 - -
          Affymetrix Rat Gene 1.0 ST Array 10727664 - -
          Affymetrix Rat Gene 1.0 ST Array 10727674 - -
          Affymetrix Rat Gene 1.0 ST Array 10727681 - -
          Affymetrix Rat Gene 1.0 ST Array 10727672 - -
          Affymetrix Rat Gene 1.1 ST Array 10727660 - -
          Rat Genome 230 2.0 Array 1367962_PM_at - -
          Rat Genome 230 2.0 Array 1367962_at - -
          Affymetrix HT Rat Focus Array 1367962_at - -
          Affymetrix HT Rat Focus Array 1367962_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0100056510.rn.1 TC0100007090.rn.1 RTA TC
          Rat Expression Set 230 1367962_at - -
          Rat Expression Set 230 1367962_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10727660 - -
          Rat Genome U34 Set rc_AI010583_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22079315 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1362652 - -
          RatRef-12 Expression BeadChip ILMN_67802 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-ffb8c786-92mpb:80/100.66.129.117:80.
          git-commit: 1a801765356e65be0b6ed8595ed79b52bb698fed
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0