Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00592373_m1
          See other TRPM4 GE Assays ›
          Gene Symbol:
          Trpm4
          Gene Name:
          transient receptor potential cation channel, subfamily M, member 4
          Gene Aliases:
          LTrpC-4, Mls2s
          Chromosome Location:
          Chr.1: 101293300 - 101323187 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00592373_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Trpm4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001136229.1 NP_001129701.1 20 - 21 3161 63
          GenBank mRNA AB040807.1 - 5 - 6 1060 63

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100005386.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          dendritic cell chemotaxis
          regulation of T cell cytokine production
          calcium ion transport
          positive regulation of cell population proliferation
          protein sumoylation
          vasoconstriction
          regulation of membrane potential
          cardiac conduction
          calcium ion transmembrane transport
          positive regulation of canonical Wnt signaling pathway
          calcium activated cation channel activity
          calcium channel activity
          calmodulin binding
          ATP binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P136072 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P369997 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 715014 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 725261 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 684422 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 861817 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 316845 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 342906 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 750689 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 290443 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 64488 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 301002 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 927234 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 606228 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 247033 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 320091 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 600001 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 207753 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 514162 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 688702 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 124751 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 258495 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 189140 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 858915 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 16899 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 90234 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 61666 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 174630 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P136072 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P369997 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P369997 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P136072 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P136072 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P369997 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P369997 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700177674 - -
          Agilent Rat cDNA Microarray (G4105A) 700177674 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10721749 - -
          Affymetrix Rat Gene 1.0 ST Array 10721764 - -
          Affymetrix Rat Gene 1.0 ST Array 10721753 - -
          Affymetrix Rat Gene 1.0 ST Array 10721746 - -
          Affymetrix Rat Gene 1.0 ST Array 10721747 - -
          Affymetrix Rat Gene 1.0 ST Array 10721755 - -
          Affymetrix Rat Gene 1.0 ST Array 10721750 - -
          Affymetrix Rat Gene 1.0 ST Array 10721751 - -
          Affymetrix Rat Gene 1.0 ST Array 10721748 - -
          Affymetrix Rat Gene 1.0 ST Array 10721756 - -
          Affymetrix Rat Gene 1.0 ST Array 10721743 - -
          Affymetrix Rat Gene 1.0 ST Array 10721752 - -
          Affymetrix Rat Gene 1.0 ST Array 10721758 - -
          Affymetrix Rat Gene 1.0 ST Array 10721763 - -
          Affymetrix Rat Gene 1.0 ST Array 10721742 - -
          Affymetrix Rat Gene 1.0 ST Array 10721759 - -
          Affymetrix Rat Gene 1.0 ST Array 10721754 - -
          Affymetrix Rat Gene 1.0 ST Array 10721744 - -
          Affymetrix Rat Gene 1.0 ST Array 10721761 - -
          Affymetrix Rat Gene 1.0 ST Array 10721740 - -
          Affymetrix Rat Gene 1.0 ST Array 10721762 - -
          Affymetrix Rat Gene 1.0 ST Array 10721757 - -
          Affymetrix Rat Gene 1.0 ST Array 10721741 - -
          Affymetrix Rat Gene 1.0 ST Array 10721745 - -
          Affymetrix Rat Gene 1.0 ST Array 10721760 - -
          Affymetrix Rat Gene 1.1 ST Array 10721739 - -
          Rat Genome 230 2.0 Array 1370392_at - -
          Rat Genome 230 2.0 Array 1370392_PM_at - -
          Affymetrix HT Rat Focus Array 1370392_at - -
          Affymetrix HT Rat Focus Array 1370392_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0100043855.rn.1 TC0100005386.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100043864.rn.1 TC0100005386.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100043850.rn.1 TC0100005386.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100043866.rn.1 TC0100005386.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100043867.rn.1 TC0100005386.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100043870.rn.1 TC0100005386.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100043875.rn.1 TC0100005386.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100043853.rn.1 TC0100005386.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100043856.rn.1 TC0100005386.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100043858.rn.1 TC0100005386.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100043849.rn.1 TC0100005386.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100043862.rn.1 TC0100005386.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100043847.rn.1 TC0100005386.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100043848.rn.1 TC0100005386.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100043851.rn.1 TC0100005386.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100043857.rn.1 TC0100005386.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100043865.rn.1 TC0100005386.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100043871.rn.1 TC0100005386.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100043873.rn.1 TC0100005386.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100043863.rn.1 TC0100005386.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100043872.rn.1 TC0100005386.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100043852.rn.1 TC0100005386.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100043854.rn.1 TC0100005386.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100043860.rn.1 TC0100005386.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100043868.rn.1 TC0100005386.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100043869.rn.1 TC0100005386.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100043859.rn.1 TC0100005386.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100043861.rn.1 TC0100005386.rn.1 RTA TC
          Rat Expression Set 230 1370392_PM_at - -
          Rat Expression Set 230 1370392_at - -
          Affymetrix Rat Gene 1.0 ST Array 10721739 - -
          Rat Genome U34 Set rc_AI234615_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21138670 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1370393 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6467d75d44-xp7n2:80/100.66.130.62:80.
          git-commit: 8ea1bebb48d33b8b7c0e2103582aa482e6db8664
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0