Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00592690_m1
          See other NLRP6 GE Assays ›
          Gene Symbol:
          Nlrp6 LOC103690037
          Gene Name:
          NLR family, pyrin domain containing 6
          NACHT, LRR and PYD domains-containing protein 6-like
          Gene Aliases:
          Avr, Nalp6, Navr, Navr/Avr
          -
          Chromosome Location:
          Chr.1: 213686017 - 213692177 on Build Rnor_6.0
          Chromosome Location:
          Chr.1: 213676328 - 213683616 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Rn00592690_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Nlrp6
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_134375.3 NP_599202.2 2121 140
          GenBank mRNA DQ631800.1 - 2121 140
          M85183.1 - 997 140
          Gene Symbol: LOC103690037
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_006230507.2 XP_006230569.2 3 - 4 3285 140

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100008059.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Nlrp6
          Click here to find siRNAs for Gene LOC103690037

          Gene Ontology Categories:

          Function(s) Process(es)

          G protein-coupled receptor signaling pathway
          response to bacterium
          regulation of autophagy
          negative regulation of toll-like receptor signaling pathway
          wound healing
          negative regulation of I-kappaB kinase/NF-kappaB signaling
          negative regulation of MAPK cascade
          regulation of inflammatory response
          negative regulation of immune response
          regulation of mucus secretion
          vasopressin receptor activity
          ATP binding
          peptide binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P023719 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 328126 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 828633 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 167856 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 813979 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 327356 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P023719 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P023719 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P023719 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P15396 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10712151 - -
          Affymetrix Rat Gene 1.0 ST Array 10712156 - -
          Affymetrix Rat Gene 1.0 ST Array 10712152 - -
          Affymetrix Rat Gene 1.0 ST Array 10712155 - -
          Affymetrix Rat Gene 1.0 ST Array 10712154 - -
          Affymetrix Rat Gene 1.0 ST Array 10712150 - -
          Affymetrix Rat Gene 1.0 ST Array 10712153 - -
          Affymetrix Rat Gene 1.1 ST Array 10712149 - -
          Rat Genome 230 2.0 Array 1369100_PM_at - -
          Rat Genome 230 2.0 Array 1369100_at - -
          Affymetrix HT Rat Focus Array 1369100_PM_at - -
          Affymetrix HT Rat Focus Array 1369100_at - -
          Affymetrix Rat Clariom S Assay PSR0100065589.rn.1 TC0100008059.rn.1 RTA TC
          Rat Expression Set 230 1369100_PM_at - -
          Rat Expression Set 230 1369100_at - -
          Affymetrix Rat Gene 1.0 ST Array 10712149 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22160377 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1366562 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-75d45cbfdf-7zrph:80/100.66.134.75:80.
          git-commit: a3c66eacce7c9fa3045440ddf4a86215532ee526
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix