Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00592850_m1
          See other ACSBG1 GE Assays ›
          Gene Symbol:
          Acsbg1
          Gene Name:
          acyl-CoA synthetase bubblegum family member 1
          Gene Aliases:
          Grlacs, Lpd
          Chromosome Location:
          Chr.8: 59184111 - 59240131 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00592850_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Acsbg1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_134389.1 NP_599216.1 8 - 9 1095 66
          XM_008766203.1 XP_008764425.1 9 - 10 1371 66
          XM_008766204.1 XP_008764426.1 9 - 10 5080 66
          GenBank mRNA AF208125.1 - 8 - 9 1095 66

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0800002581.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Acsbg1

          Gene Ontology Categories:

          Function(s) Process(es)

          very long-chain fatty acid metabolic process
          ovarian follicle atresia
          long-chain fatty acid metabolic process
          response to glucocorticoid
          long-chain fatty acid-CoA ligase activity
          ATP binding
          very long-chain fatty acid-CoA ligase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P13257 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 969119 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 240219 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 455922 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 571135 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 283378 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 568719 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 623091 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 479557 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 671370 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 670026 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 367075 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 78486 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 846198 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 666339 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 545503 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 54710 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 378545 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 544860 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 145626 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 306598 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 503482 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 810527 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 917001 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 730911 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 770305 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 429325 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 679481 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 909893 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 346056 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 641557 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 921073 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 97319 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 106986 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 736268 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 292425 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 955842 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 269408 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 789782 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 653737 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 928203 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 25404 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 102242 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 127920 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 55769 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 127461 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 101339 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 119157 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P13257 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P13257 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P13257 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P13257 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 84696 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 104130 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 16708 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10917575 - -
          Affymetrix Rat Gene 1.0 ST Array 10917581 - -
          Affymetrix Rat Gene 1.0 ST Array 10917582 - -
          Affymetrix Rat Gene 1.0 ST Array 10917571 - -
          Affymetrix Rat Gene 1.0 ST Array 10917577 - -
          Affymetrix Rat Gene 1.0 ST Array 10917572 - -
          Affymetrix Rat Gene 1.0 ST Array 10917573 - -
          Affymetrix Rat Gene 1.0 ST Array 10917570 - -
          Affymetrix Rat Gene 1.0 ST Array 10917574 - -
          Affymetrix Rat Gene 1.0 ST Array 10917579 - -
          Affymetrix Rat Gene 1.0 ST Array 10917569 - -
          Affymetrix Rat Gene 1.0 ST Array 10917576 - -
          Affymetrix Rat Gene 1.0 ST Array 10917580 - -
          Affymetrix Rat Gene 1.0 ST Array 10917578 - -
          Affymetrix Rat Gene 1.1 ST Array 10917568 - -
          Rat Genome 230 2.0 Array 1367923_PM_at - -
          Rat Genome 230 2.0 Array 1367923_at - -
          Affymetrix HT Rat Focus Array 1367923_at - -
          Affymetrix HT Rat Focus Array 1367923_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0800020866.rn.1 TC0800002581.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800020868.rn.1 TC0800002581.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800020871.rn.1 TC0800002581.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800020861.rn.1 TC0800002581.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800020855.rn.1 TC0800002581.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800020856.rn.1 TC0800002581.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800020858.rn.1 TC0800002581.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800020870.rn.1 TC0800002581.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800020873.rn.1 TC0800002581.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800020872.rn.1 TC0800002581.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800020857.rn.1 TC0800002581.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800020867.rn.1 TC0800002581.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800020862.rn.1 TC0800002581.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800020869.rn.1 TC0800002581.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800020859.rn.1 TC0800002581.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800020864.rn.1 TC0800002581.rn.1 RTA TC
          Rat Expression Set 230 1367923_at - -
          Rat Expression Set 230 1367923_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10917568 - -
          Rat Genome U34 Set rc_AI101794_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21252277 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1370682 - -
          RatRef-12 Expression BeadChip ILMN_61941 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0