Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00592866_m1
          See other SPATA6 GE Assays ›
          Gene Symbol:
          Spata6
          Gene Name:
          spermatogenesis associated 6
          Gene Aliases:
          Hash
          Chromosome Location:
          Chr.5: 131719922 - 131803853 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00592866_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Spata6
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_134392.2 NP_599219.2 8 - 9 1075 67
          GenBank mRNA AB046606.1 - 1 - 2 96 67
          AF291466.1 - 8 - 9 1291 67
          BC126081.1 - 8 - 9 1008 67
          BC158581.1 - 8 - 9 1024 67

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0500001210.rn.1

          Additional Information:

          This assay is in the 5'UTR region of the AB046606.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Spata6

          Gene Ontology Categories:

          Function(s)

          multicellular organism development
          spermatogenesis
          cell differentiation

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P323280 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 936075 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 666533 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 310134 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 403785 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 526229 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 930448 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 401949 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 606467 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 527272 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 640859 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 810545 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 549075 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 603044 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 805174 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 597507 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 863224 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 278137 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 477819 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 111645 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 508756 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 667310 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 491439 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 946205 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 127830 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 243905 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 644071 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 613200 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 217473 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 283691 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 432309 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 2046 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 214092 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 961588 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 380695 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 394515 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 479946 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 856642 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 215309 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 922642 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 675578 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 62027 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 687648 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 51703 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 434250 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 829126 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 314026 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 626643 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 219856 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 286195 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 36689 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 267431 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 479637 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 24116 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 799443 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 663368 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 138499 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 99586 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 125510 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 63354 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 12017 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 73886 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 122865 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 161385 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 158924 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 59893 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 138076 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 124675 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P323280 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P323280 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P323280 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P182182 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P323280 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 22034 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 102568 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 85397 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 97568 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 20554 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 6244 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10871015 - -
          Affymetrix Rat Gene 1.0 ST Array 10871011 - -
          Affymetrix Rat Gene 1.0 ST Array 10871008 - -
          Affymetrix Rat Gene 1.0 ST Array 10871009 - -
          Affymetrix Rat Gene 1.0 ST Array 10871006 - -
          Affymetrix Rat Gene 1.0 ST Array 10871013 - -
          Affymetrix Rat Gene 1.0 ST Array 10871019 - -
          Affymetrix Rat Gene 1.0 ST Array 10871017 - -
          Affymetrix Rat Gene 1.0 ST Array 10871016 - -
          Affymetrix Rat Gene 1.0 ST Array 10871014 - -
          Affymetrix Rat Gene 1.0 ST Array 10871018 - -
          Affymetrix Rat Gene 1.0 ST Array 10871005 - -
          Affymetrix Rat Gene 1.0 ST Array 10871010 - -
          Affymetrix Rat Gene 1.0 ST Array 10871020 - -
          Affymetrix Rat Gene 1.0 ST Array 10871007 - -
          Affymetrix Rat Gene 1.1 ST Array 10871004 - -
          Rat Genome 230 2.0 Array 1368644_PM_at - -
          Rat Genome 230 2.0 Array 1368644_at - -
          Rat Genome 230 2.0 Array 1368643_at - -
          Rat Genome 230 2.0 Array 1368643_PM_at - -
          Affymetrix HT Rat Focus Array 1368644_at - -
          Affymetrix HT Rat Focus Array 1368644_PM_at - -
          Affymetrix HT Rat Focus Array 1368643_at - -
          Affymetrix HT Rat Focus Array 1368643_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0500008253.rn.1 TC0500001210.rn.1 RTA TC
          Rat Expression Set 230 1368644_PM_at - -
          Rat Expression Set 230 1368643_at - -
          Rat Expression Set 230 1368643_PM_at - -
          Rat Expression Set 230 1368644_at - -
          Affymetrix Rat Gene 1.0 ST Array 10871004 - -
          Rat Genome U34 Set rc_AI233848_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22363055 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376570 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-ffb8c786-92mpb:80/100.66.129.117:80.
          git-commit: 1a801765356e65be0b6ed8595ed79b52bb698fed
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0