Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00594023_m1
          See other SMAD9 GE Assays ›
          Gene Symbol:
          Smad9 LOC103691556
          Gene Name:
          SMAD family member 9
          mothers against decapentaplegic homolog 9
          Gene Aliases:
          Madh9, Smad8
          -
          Chromosome Location:
          Chr.2: 143981134 - 143998165 on Build Rnor_6.0
          Chromosome Location:
          Chr.2: 144191095 - 144211986 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Rn00594023_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Smad9
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_138872.1 NP_620227.1 819 66
          XM_008761009.1 XP_008759231.1 2 - 3 1614 66
          GenBank mRNA AF012347.1 - 819 66
          Gene Symbol: LOC103691556
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_008761019.1 XP_008759241.1 1614 66
          XM_008761020.1 XP_008759242.1 1614 66

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0200001161.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Smad9
          Click here to find siRNAs for Gene LOC103691556

          Gene Ontology Categories:

          Function(s) Process(es)

          ureteric bud development
          response to hypoxia
          Mullerian duct regression
          transcription, DNA-templated
          protein phosphorylation
          transforming growth factor beta receptor signaling pathway
          BMP signaling pathway
          midbrain development
          hindbrain development
          positive regulation of cell differentiation
          positive regulation of transcription, DNA-templated
          cartilage development
          bone development
          cellular response to organic cyclic compound
          DNA binding
          DNA-binding transcription factor activity
          protein binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P068197 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 474967 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 314091 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 32330 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 318776 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 39804 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 4249 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 86650 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 99335 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 832861 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 785 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 87550 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 118017 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P068197 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P068197 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P068197 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P545792 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 66856 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 17360 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10815438 - -
          Affymetrix Rat Gene 1.0 ST Array 10815441 - -
          Affymetrix Rat Gene 1.0 ST Array 10815437 - -
          Affymetrix Rat Gene 1.0 ST Array 10815439 - -
          Affymetrix Rat Gene 1.0 ST Array 10815440 - -
          Affymetrix Rat Gene 1.1 ST Array 10815436 - -
          Rat Genome 230 2.0 Array 1370716_PM_at - -
          Rat Genome 230 2.0 Array 1370716_at - -
          Affymetrix HT Rat Focus Array 1370716_PM_at - -
          Affymetrix HT Rat Focus Array 1370716_at - -
          Affymetrix Rat Clariom S Assay PSR0200007344.rn.1 TC0200001161.rn.1 RTA TC
          Rat Expression Set 230 1370716_PM_at - -
          Rat Expression Set 230 1370716_at - -
          Affymetrix Rat Gene 1.0 ST Array 10815436 - -
          Rat Genome U34 Set AF012347_at - -
          Rat Genome U34 Set AF012347_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21778762 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1362558 - -
          RatRef-12 Expression BeadChip ILMN_69209 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-9d5985c9f-cd4wl:80/100.66.130.62:80.
          git-commit: 2310f86ea667df8f532fad8d69a2d0b8b05e2a1e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0