Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00594782_m1
          See other COMMD5 GE Assays ›
          Gene Symbol:
          Commd5 LOC100910472
          Gene Name:
          COMM domain containing 5
          COMM domain-containing protein 5-like
          Gene Aliases:
          Hcarg
          -
          Chromosome Location:
          Chr.7: 118502349 - 118504416 on Build Rnor_6.0
          Chromosome Location:
          Chr.7: 117962943 - 117965010 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Rn00594782_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Commd5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_139108.2 NP_620808.1 69 61
          GenBank mRNA AF290194.1 - 74 61
          Gene Symbol: LOC100910472
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_006241916.2 XP_006241978.1 1 - 2 398 61

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the AF290194.1, NM_139108.2, XM_006241916.2 transcripts.
          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Commd5
          Click here to find siRNAs for Gene LOC100910472

          Gene Ontology Categories:

          Function(s)

          regulation of cell growth
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          regulation of cell differentiation
          proximal tubule morphogenesis

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P308673 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 721428 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 685668 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P308673 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P308673 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P308673 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P308673 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700246243 - -
          Agilent Rat cDNA Microarray (G4105A) 700246243 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10897395 - -
          Affymetrix Rat Gene 1.0 ST Array 10897396 - -
          Affymetrix Rat Gene 1.1 ST Array 10897394 - -
          Rat Genome 230 2.0 Array 1370327_at - -
          Rat Genome 230 2.0 Array 1370327_PM_at - -
          Affymetrix HT Rat Focus Array 1370327_at - -
          Affymetrix HT Rat Focus Array 1370327_PM_at - -
          Rat Expression Set 230 1370327_PM_at - -
          Rat Expression Set 230 1370327_at - -
          Affymetrix Rat Gene 1.0 ST Array 10897394 - -
          Rat Genome U34 Set rc_AA899199_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21270888 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1349253 - -
          RatRef-12 Expression BeadChip ILMN_63265 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-865d7754c7-mcq8j:80/100.66.130.62:80.
          git-commit: 7ff36f61b459eb8f02ba5a263f0e3ce367ca82d3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0