Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00594894_g1
          See other SCD1 GE Assays ›
          Gene Symbol:
          Scd1
          Gene Name:
          stearoyl-Coenzyme A desaturase 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 264159966 - 264173061 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00594894_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information
          • Citations (2)

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Scd1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_139192.2 NP_631931.2 1 - 2 71 86
          XM_006231433.2 XP_006231495.1 2 - 3 206 86
          GenBank mRNA AF509568.1 - 1 - 2 71 86
          AF509569.1 - 1 - 2 71 86
          J02585.1 - 1 - 2 125 86

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the AF509568.1, AF509569.1, J02585.1, NM_139192.2, XM_006231433.2 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Detect Genomic DNA Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Scd1

          Gene Ontology Categories:

          Function(s) Process(es)

          fatty acid biosynthetic process
          lipid biosynthetic process
          cholesterol esterification
          negative regulation of growth of symbiont in host
          defense response to Gram-positive bacterium
          white fat cell differentiation
          brown fat cell differentiation
          oxidation-reduction process
          stearoyl-CoA 9-desaturase activity
          iron ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P085530 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 360006 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 860800 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 244846 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 782831 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 649765 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 73965 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 934590 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 517108 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 459784 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 166601 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P085530 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P085530 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P085530 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P550145 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700330140 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 27565 - -
          Agilent Rat cDNA Microarray (G4105A) 700330140 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10730350 - -
          Affymetrix Rat Gene 1.0 ST Array 10730355 - -
          Affymetrix Rat Gene 1.0 ST Array 10730354 - -
          Affymetrix Rat Gene 1.0 ST Array 10730352 - -
          Affymetrix Rat Gene 1.0 ST Array 10730351 - -
          Affymetrix Rat Gene 1.1 ST Array 10730349 - -
          Rat Genome 230 2.0 Array 1370355_at - -
          Rat Genome 230 2.0 Array 1370355_PM_at - -
          Affymetrix HT Rat Focus Array 1370355_PM_at - -
          Affymetrix HT Rat Focus Array 1370355_at - -
          Affymetrix Rat Clariom S Assay PSR0100062603.rn.1 TC0100007769.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100062597.rn.1 TC0100007769.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100062601.rn.1 TC0100007769.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100062599.rn.1 TC0100007769.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100062602.rn.1 TC0100007769.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100062598.rn.1 TC0100007769.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100062600.rn.1 TC0100007769.rn.1 RTA TC
          Rat Expression Set 230 1370355_at - -
          Rat Expression Set 230 1370355_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10730349 - -
          Rat Genome U34 Set AF036761_g_at - -
          Rat Genome U34 Set rc_AI175764_s_at - -
          Rat Genome U34 Set J02585_at - -
          Rat Genome U34 Set J02585_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21664251 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1374191 - -
          RatRef-12 Expression BeadChip ILMN_59561 - -

          Back To Top
           

          Citations (2)

          Causes and prevention of tamoxifen-induced accumulation of triacylglycerol in rat liver.
          Authors
          Gudbrandsen OA, Rost TH, Berge RK
          Journal
          J Lipid Res - Page No 2223 (Sun Oct 01 00:00:00 EDT 2006)
          Volume (Year)
          47 (2006)
          Product Usage
          -
          Differential activation of ER stress and apoptosis in response to chronically elevated free fatty acids in pancreatic beta-cells.
          Authors
          Lai E, Bikopoulos G, Wheeler MB, Rozakis-Adcock M, Volchuk A
          Journal
          Am J Physiol Endocrinol Metab - Page No E540 (Sat Mar 01 00:00:00 EST 2008)
          Volume (Year)
          294 (2008)
          Product Usage
          -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-786bfc498d-47c8f:80/100.66.130.62:80.
          git-commit: 9eb01563f6748445c58f67612c67ecf6b1da7421
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0