Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00596219_m1
          See other SFXN5 GE Assays ›
          Gene Symbol:
          Sfxn5
          Gene Name:
          sideroflexin 5
          Gene Aliases:
          BBG-TCC
          Chromosome Location:
          Chr.4: 116992645 - 117112141 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00596219_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Sfxn5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_153298.1 NP_695210.1 1 - 2 148 95
          XM_008763051.1 XP_008761273.1 1 - 2 741 95
          GenBank mRNA AB056724.1 - 1 - 2 165 95

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0400003212.rn.1

          Additional Information:

          This assay is in the 5'UTR region of the XM_008763051.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Sfxn5

          Gene Ontology Categories:

          Function(s) Process(es)

          citrate transport
          ion transmembrane transport
          iron ion homeostasis
          citrate transmembrane transporter activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P200846 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 823050 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 953661 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 138514 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 165167 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 827072 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 658005 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 707707 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 465104 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 930068 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 426270 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 527090 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 181326 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 442650 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 210236 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 640645 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 247250 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 968023 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 678967 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 266208 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 653116 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 492592 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 19889 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 504129 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 84138 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 16051 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 137681 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 892688 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 429089 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 386173 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 66312 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 287757 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 382914 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 455521 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 279625 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 88944 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 959300 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 193600 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 760812 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 765309 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 246373 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 389725 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 964873 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 900362 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 482696 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 578942 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 147881 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 189949 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 897188 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 290467 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 233967 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 155016 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 793380 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 603863 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 409964 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 622043 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 935708 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 412578 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 250783 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 708833 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 129142 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 517519 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 655462 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 492761 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 154446 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 816663 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 610250 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 267230 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 250752 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 643648 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 79855 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 137900 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 898467 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 36169 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 904021 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 309904 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 117779 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 284243 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 559388 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 672117 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 424547 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 555412 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 797469 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 130228 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 173046 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 10783 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 146338 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 162998 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 166300 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 149287 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 62797 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 72864 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 174537 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 95110 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 57454 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 44474 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 107421 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P200846 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P200846 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P200846 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P200846 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 46698 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 15270 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 85055 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 58334 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 33049 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 71019 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 53162 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 63327 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10863647 - -
          Affymetrix Rat Gene 1.0 ST Array 10863643 - -
          Affymetrix Rat Gene 1.0 ST Array 10863645 - -
          Affymetrix Rat Gene 1.0 ST Array 10863640 - -
          Affymetrix Rat Gene 1.0 ST Array 10863648 - -
          Affymetrix Rat Gene 1.0 ST Array 10863642 - -
          Affymetrix Rat Gene 1.0 ST Array 10863641 - -
          Affymetrix Rat Gene 1.0 ST Array 10863644 - -
          Affymetrix Rat Gene 1.0 ST Array 10863646 - -
          Affymetrix Rat Gene 1.1 ST Array 10863639 - -
          Rat Genome 230 2.0 Array 1389491_PM_at - -
          Rat Genome 230 2.0 Array 1375621_PM_at - -
          Rat Genome 230 2.0 Array 1375621_at - -
          Rat Genome 230 2.0 Array 1389491_at - -
          Affymetrix HT Rat Focus Array 1375621_PM_at - -
          Affymetrix HT Rat Focus Array 1375621_at - -
          Affymetrix Rat Clariom S Assay PSR0400022854.rn.1 TC0400003212.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400022853.rn.1 TC0400003212.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400022852.rn.1 TC0400003212.rn.1 RTA TC
          Rat Expression Set 230 1375621_PM_at - -
          Rat Expression Set 230 1375621_at - -
          Affymetrix Rat Gene 1.0 ST Array 10863639 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21520828 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1353083 - -
          RatRef-12 Expression BeadChip ILMN_66383 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7b745768ff-vzrjf:80/100.66.128.131:80.
          git-commit: 4b95038805a43672018eef8c57cc3f15d8b0c074
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: integration/2.33.0-meta-desc