Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00597012_m1
          See other LOC100912430 GE Assays ›
          Gene Symbol:
          LOC100912430 Gstm5
          Gene Name:
          glutathione S-transferase Mu 5-like
          glutathione S-transferase, mu 5
          Gene Aliases:
          -
          -
          Chromosome Location:
          Chr.Un|NW_007906853: 52914 - 55985 on Build Rnor_6.0
          Chromosome Location:
          Chr.2: 210685338 - 210688133 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Rn00597012_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC100912430
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_003749378.3 XP_003749426.1 5 - 6 511 68
          Gene Symbol: Gstm5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_172038.1 NP_742035.1 5 - 6 375 68
          GenBank mRNA U86635.1 - 5 - 6 375 68

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene LOC100912430
          Click here to find siRNAs for Gene Gstm5

          Gene Ontology Categories:

          Function(s) Process(es)

          glutathione metabolic process
          nitrobenzene metabolic process
          xenobiotic catabolic process
          response to estrogen
          cellular detoxification of nitrogen compound
          biological_process
          glutathione transferase activity
          enzyme binding
          identical protein binding
          protein homodimerization activity
          glutathione binding
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P331464 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 472647 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 710006 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P331464 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P331464 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P331464 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P517303 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P331464 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P517296 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10818295 - -
          Affymetrix Rat Gene 1.0 ST Array 10818292 - -
          Affymetrix Rat Gene 1.0 ST Array 10818294 - -
          Affymetrix Rat Gene 1.0 ST Array 10818298 - -
          Affymetrix Rat Gene 1.0 ST Array 10818296 - -
          Affymetrix Rat Gene 1.0 ST Array 10818299 - -
          Affymetrix Rat Gene 1.0 ST Array 10818300 - -
          Affymetrix Rat Gene 1.0 ST Array 10818301 - -
          Affymetrix Rat Gene 1.0 ST Array 10818293 - -
          Affymetrix Rat Gene 1.0 ST Array 10818297 - -
          Affymetrix Rat Gene 1.1 ST Array 10818291 - -
          Rat Genome 230 2.0 Array 1370813_at - -
          Rat Genome 230 2.0 Array 1370813_PM_at - -
          Affymetrix HT Rat Focus Array 1370813_at - -
          Affymetrix HT Rat Focus Array 1370813_PM_at - -
          Rat Expression Set 230 1370813_PM_at - -
          Rat Expression Set 230 1370813_at - -
          Affymetrix Rat Gene 1.0 ST Array 10818291 - -
          Rat Genome U34 Set U86635_at - -
          Rat Genome U34 Set U86635_g_at - -
          Rat Genome U34 Set rc_AA957007_at - -
          Rat Genome U34 Set U86635_g_at - -
          Rat Genome U34 Set U86635_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21563029 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1352924 - -
          RatRef-12 Expression BeadChip ILMN_62634 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0