Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00597492_m1
          See other NPC2 GE Assays ›
          Gene Symbol:
          Npc2
          Gene Name:
          Niemann-Pick disease, type C2
          Gene Aliases:
          re1
          Chromosome Location:
          Chr.6: 108467410 - 108488330 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00597492_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Npc2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_173118.2 NP_775141.2 1 - 2 185 68
          GenBank mRNA AJ515237.1 - 1 - 2 120 68
          BC058132.1 - 1 - 2 148 68
          FQ214040.1 - 1 - 2 106 68
          FQ216545.1 - 1 - 2 110 68
          FQ219301.1 - 1 - 2 127 68
          FQ219863.1 - 1 - 2 118 68
          FQ220660.1 - 1 - 2 106 68
          FQ220919.1 - 1 - 2 112 68
          FQ228456.1 - 1 - 2 110 68
          FQ229422.1 - 1 - 2 165 68
          FQ229782.1 - 1 - 2 106 68
          FQ229783.1 - 1 - 2 108 68
          FQ230107.1 - 1 - 2 112 68

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Npc2

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          response to virus
          cholesterol transport
          intracellular sterol transport
          intracellular cholesterol transport
          cholesterol efflux
          cholesterol homeostasis
          molecular_function
          cholesterol binding
          enzyme binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P139742 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 507339 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 805623 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 468358 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 192789 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 956825 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 588586 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 581154 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 540129 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 191753 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 835160 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 121814 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 30850 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 806312 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 557156 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 564885 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 924028 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 51652 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 17392 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 82145 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P139742 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P139742 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P139742 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P139742 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 64493 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 10188 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10891164 - -
          Affymetrix Rat Gene 1.0 ST Array 10891162 - -
          Affymetrix Rat Gene 1.0 ST Array 10891163 - -
          Affymetrix Rat Gene 1.0 ST Array 10891161 - -
          Affymetrix Rat Gene 1.1 ST Array 10891160 - -
          Rat Genome 230 2.0 Array 1398892_at - -
          Rat Genome 230 2.0 Array 1398892_PM_at - -
          Affymetrix HT Rat Focus Array 1398892_at - -
          Affymetrix HT Rat Focus Array 1398892_PM_at - -
          Rat Expression Set 230 1398892_at - -
          Rat Expression Set 230 1398892_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10891160 - -
          Rat Genome U34 Set rc_AI231785_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22176686 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1352122 - -
          RatRef-12 Expression BeadChip ILMN_59161 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5777fb6d44-4pg6x:80/100.66.134.75:80.
          git-commit: 3a4c0ffdaddae765c0fdfab5c280296089abf14d
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0