Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00678044_g1
          See other BIK GE Assays ›
          Gene Symbol:
          Bik
          Gene Name:
          BCL2-interacting killer (apoptosis-inducing)
          Gene Aliases:
          Biklk, Blk
          Chromosome Location:
          Chr.7: 124390926 - 124410449 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00678044_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Bik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053704.1 NP_446156.1 4 - 5 491 79
          XM_006242048.2 XP_006242110.1 4 - 5 578 79
          GenBank mRNA AF372501.1 - 4 - 5 491 79
          FQ225338.1 - 4 - 5 468 79

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0700001550.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Detect Genomic DNA Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Bik

          Gene Ontology Categories:

          Function(s) Process(es)

          spermatogenesis
          male gonad development
          apoptotic mitochondrial changes
          positive regulation of protein homooligomerization
          positive regulation of release of cytochrome c from mitochondria
          protein heterodimerization activity
          BH domain binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P12905 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 798072 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 810438 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 527680 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 909526 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 652972 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 225755 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 915602 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 900251 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 720424 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 371496 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 587316 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 45151 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 519175 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 675543 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 379135 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 147404 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 153983 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 58291 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P12905 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P12905 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P12905 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P12905 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 93136 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 92253 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10905824 - -
          Affymetrix Rat Gene 1.0 ST Array 10905826 - -
          Affymetrix Rat Gene 1.0 ST Array 10905825 - -
          Affymetrix Rat Gene 1.0 ST Array 10905823 - -
          Affymetrix Rat Gene 1.1 ST Array 10905822 - -
          Rat Genome 230 2.0 Array 1387249_at - -
          Rat Genome 230 2.0 Array 1387249_PM_at - -
          Affymetrix HT Rat Focus Array 1387249_at - -
          Affymetrix HT Rat Focus Array 1387249_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0700011804.rn.1 TC0700001550.rn.1 RTA TC
          Rat Expression Set 230 1387249_at - -
          Rat Expression Set 230 1387249_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10905822 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20747619 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1365163 - -
          RatRef-12 Expression BeadChip ILMN_68320 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7699c4d8ff-7jgns:80/100.66.130.62:80.
          git-commit: c11338b6b8d815146c5ef25758b63739976b60cb
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline