Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00678055_g1
          See other HDGF GE Assays ›
          Gene Symbol:
          Hdgf
          Gene Name:
          hepatoma-derived growth factor
          Gene Aliases:
          -
          Chromosome Location:
          Chr.2: 187275120 - 187284310 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00678055_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Hdgf
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053707.2 NP_446159.1 4 - 5 760 74
          GenBank mRNA AF389348.1 - 4 - 5 489 74
          AF448810.1 - 4 - 5 489 74
          BC070943.1 - 4 - 5 760 74
          FQ231256.1 - 5 - 6 523 74

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0200001519.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Hdgf

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription by RNA polymerase II
          transcription, DNA-templated
          cellular process
          protein localization to nucleus
          positive regulation of transcription by RNA polymerase II
          nucleotide binding
          RNA polymerase II cis-regulatory region sequence-specific DNA binding
          transcription corepressor binding
          transcription corepressor activity
          RNA binding
          growth factor activity
          heparin binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P009375 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P336429 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 578949 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 942423 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 541324 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 371609 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 754094 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 577018 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 46084 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P336429 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P009375 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P009375 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P336429 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P009375 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P336429 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P336429 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 103006 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10816389 - -
          Affymetrix Rat Gene 1.0 ST Array 10816393 - -
          Affymetrix Rat Gene 1.0 ST Array 10816392 - -
          Affymetrix Rat Gene 1.0 ST Array 10816390 - -
          Affymetrix Rat Gene 1.0 ST Array 10816391 - -
          Affymetrix Rat Gene 1.0 ST Array 10816388 - -
          Affymetrix Rat Gene 1.1 ST Array 10816387 - -
          Rat Genome 230 2.0 Array 1367817_at - -
          Rat Genome 230 2.0 Array 1367817_PM_at - -
          Affymetrix HT Rat Focus Array 1367817_at - -
          Affymetrix HT Rat Focus Array 1367817_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0200009403.rn.1 TC0200001519.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200009407.rn.1 TC0200001519.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200009409.rn.1 TC0200001519.rn.1 RTA TC
          Rat Expression Set 230 1367817_at - -
          Rat Expression Set 230 1367817_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10816387 - -
          Rat Genome U34 Set rc_AI170366_at - -
          Rat Genome U34 Set rc_AI180266_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21641475 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1358138 - -
          RatRef-12 Expression BeadChip ILMN_66360 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0