Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00679622_m1
          See other RUNX3 GE Assays ›
          Gene Symbol:
          Runx3
          Gene Name:
          runt-related transcription factor 3
          Gene Aliases:
          -
          Chromosome Location:
          Chr.5: 153507093 - 153531137 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00679622_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Runx3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_130425.1 NP_569109.1 4 - 5 694 103
          GenBank mRNA AF421886.1 - 4 - 5 694 103
          FQ230848.1 - 443 103

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0500001673.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Rattus norvegicus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Runx3

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription by RNA polymerase II
          chondrocyte differentiation
          regulation of transcription, DNA-templated
          protein phosphorylation
          axon guidance
          biological_process
          hair follicle morphogenesis
          neuron projection development
          interferon-gamma production
          mRNA transcription by RNA polymerase II
          negative regulation of cell cycle
          cell maturation
          negative regulation of epithelial cell proliferation
          positive regulation of extrinsic apoptotic signaling pathway
          RNA polymerase II transcription regulatory region sequence-specific DNA binding
          DNA-binding transcription factor activity, RNA polymerase II-specific
          molecular_function
          DNA binding
          DNA-binding transcription factor activity
          ATP binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P051318 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 199263 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 827821 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 801794 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 497708 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 182083 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 798032 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 125931 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 722566 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 957831 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 555638 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 200230 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 413449 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 346637 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 897071 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 838124 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 116891 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 72217 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 717204 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 819572 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 18911 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 167472 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 141774 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P051317 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P051317 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P051317 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P13119 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 8148 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 53459 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10872848 - -
          Affymetrix Rat Gene 1.0 ST Array 10872849 - -
          Affymetrix Rat Gene 1.0 ST Array 10872846 - -
          Affymetrix Rat Gene 1.0 ST Array 10872847 - -
          Affymetrix Rat Gene 1.0 ST Array 10872845 - -
          Affymetrix Rat Gene 1.1 ST Array 10872843 - -
          Rat Genome 230 2.0 Array 1390946_PM_at - -
          Rat Genome 230 2.0 Array 1390946_at - -
          Rat Genome 230 2.0 Array 1387479_PM_at - -
          Rat Genome 230 2.0 Array 1387479_at - -
          Affymetrix HT Rat Focus Array 1387479_PM_at - -
          Affymetrix HT Rat Focus Array 1387479_at - -
          Affymetrix Rat Clariom S Assay PSR0500012085.rn.1 TC0500001673.rn.1 RTA TC
          Rat Expression Set 230 1387479_at - -
          Rat Expression Set 230 1387479_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10872843 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21661582 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1372259 - -
          RatRef-12 Expression BeadChip ILMN_54863 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-66756dbf47-27kf9:80/100.66.130.62:80.
          git-commit: ed967532dff6834307dac93af002e5c8e22ba7c9
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline