Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00680393_m1
          See other CD53 GE Assays ›
          Gene Symbol:
          Cd53
          Gene Name:
          Cd53 molecule
          Gene Aliases:
          OX44, Ox-44
          Chromosome Location:
          Chr.2: 209489279 - 209537098 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00680393_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cd53
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_012523.2 NP_036655.1 5 - 6 565 61
          GenBank mRNA BC078900.1 - 5 - 6 516 61
          M57276.1 - 5 - 6 579 61

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0200004423.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cd53

          Gene Ontology Categories:

          Function(s)

          regulation of growth
          regulation of myoblast differentiation
          cell-cell adhesion
          positive regulation of myoblast fusion

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P120941 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 615889 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 25848 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 624996 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 545286 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 599535 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 835404 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 390971 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 681266 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 45844 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 44129 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 572868 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 181868 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 109049 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 63154 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 955886 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 547069 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 81031 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 106010 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 60886 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 116438 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 13494 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 31145 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 84500 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P120941 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P120941 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P120941 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 53488 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 10935 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 1782 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 26067 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10825813 - -
          Affymetrix Rat Gene 1.0 ST Array 10825810 - -
          Affymetrix Rat Gene 1.0 ST Array 10825812 - -
          Affymetrix Rat Gene 1.0 ST Array 10825811 - -
          Affymetrix Rat Gene 1.1 ST Array 10825809 - -
          Rat Genome 230 2.0 Array 1368518_PM_at - -
          Rat Genome 230 2.0 Array 1368518_at - -
          Affymetrix HT Rat Focus Array 1368518_PM_at - -
          Affymetrix HT Rat Focus Array 1368518_at - -
          Affymetrix Rat Clariom S Assay PSR0200030569.rn.1 TC0200004423.rn.1 RTA TC
          Rat Expression Set 230 1368518_at - -
          Rat Expression Set 230 1368518_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10825809 - -
          Rat Genome U34 Set M57276_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21933940 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1353693 - -
          RatRef-12 Expression BeadChip ILMN_58541 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0