Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00680664_g1
          See other PSMB9 GE Assays ›
          Gene Symbol:
          Psmb9
          Gene Name:
          proteasome (prosome, macropain) subunit, beta type, 9
          Gene Aliases:
          Lmp2
          Chromosome Location:
          Chr.20: 3973424 - 3978845 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00680664_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Psmb9
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_012708.2 NP_036840.2 4 - 5 441 88
          GenBank mRNA BC091161.1 - 4 - 5 429 88
          BP500440.1 - 4 - 5 446 88
          D10757.1 - 4 - 5 408 88
          FQ228039.1 - 4 - 5 415 88
          KC222887.1 - 4 - 5 390 88
          KC222888.1 - 4 - 5 390 88
          KC222889.1 - 4 - 5 390 88
          KC222890.1 - 4 - 5 390 88
          KC222891.1 - 4 - 5 390 88

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC2000001061.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Psmb9

          Gene Ontology Categories:

          Function(s) Process(es)

          immune system process
          antigen processing and presentation
          proteolysis involved in cellular protein catabolic process
          threonine-type endopeptidase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P301963 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 340910 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 577742 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 647688 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 327026 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 157889 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P301963 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P301963 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P301963 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P301963 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700542577 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 53347 - -
          Agilent Rat cDNA Microarray (G4105A) 700542577 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10828359 - -
          Affymetrix Rat Gene 1.0 ST Array 10828363 - -
          Affymetrix Rat Gene 1.0 ST Array 10828362 - -
          Affymetrix Rat Gene 1.0 ST Array 10828358 - -
          Affymetrix Rat Gene 1.0 ST Array 10828361 - -
          Affymetrix Rat Gene 1.0 ST Array 10828360 - -
          Affymetrix Rat Gene 1.1 ST Array 10828357 - -
          Rat Genome 230 2.0 Array 1370186_PM_at - -
          Rat Genome 230 2.0 Array 1370186_at - -
          Affymetrix HT Rat Focus Array 1370186_at - -
          Affymetrix HT Rat Focus Array 1370186_PM_at - -
          Affymetrix Rat Clariom S Assay PSR2000008639.rn.1 TC2000001061.rn.1 RTA TC
          Rat Expression Set 230 1370186_PM_at - -
          Rat Expression Set 230 1370186_at - -
          Affymetrix Rat Gene 1.0 ST Array 10828357 - -
          Rat Genome U34 Set D10757_g_at - -
          Rat Genome U34 Set D10757_at - -
          Rat Genome U34 Set rc_AI012340_s_at - -
          Rat Genome U34 Set rc_AI177481_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21903077 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1350444 - -
          RatRef-12 Expression BeadChip ILMN_61925 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0