Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00680751_g1
          See other COQ7 GE Assays ›
          Gene Symbol:
          Coq7
          Gene Name:
          coenzyme Q7 homolog, ubiquinone (yeast)
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 188176060 - 188190874 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00680751_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Coq7
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_012785.3 NP_036917.2 5 - 6 640 68
          XM_006230108.2 XP_006230170.1 5 - 6 714 68
          GenBank mRNA AI711299.1 - 1 - 2 68
          BF558335.1 - 1 - 2 68
          FQ225865.1 - 640 68
          U46149.1 - 4 - 5 466 68

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100006494.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Coq7

          Gene Ontology Categories:

          Function(s) Process(es)

          age-dependent response to oxidative stress
          in utero embryonic development
          neural tube formation
          ubiquinone biosynthetic process
          response to oxidative stress
          determination of adult lifespan
          response to xenobiotic stimulus
          neurogenesis
          respiratory electron transport chain
          cellular response to oxidative stress
          mitochondrial ATP synthesis coupled electron transport
          mitochondrion morphogenesis
          molecular_function
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P505282 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 937778 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 927746 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 775572 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 57827 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 911699 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 154293 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P505282 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P505282 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P505282 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P505282 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700935149 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 31134 - -
          Agilent Rat cDNA Microarray (G4105A) 700935149 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10725248 - -
          Affymetrix Rat Gene 1.0 ST Array 10725249 - -
          Affymetrix Rat Gene 1.0 ST Array 10725247 - -
          Affymetrix Rat Gene 1.0 ST Array 10725246 - -
          Affymetrix Rat Gene 1.0 ST Array 10725250 - -
          Affymetrix Rat Gene 1.1 ST Array 10725245 - -
          Rat Genome 230 2.0 Array 1370506_at - -
          Rat Genome 230 2.0 Array 1370506_PM_at - -
          Affymetrix HT Rat Focus Array 1370506_PM_at - -
          Affymetrix HT Rat Focus Array 1370506_at - -
          Affymetrix Rat Clariom S Assay PSR0100050979.rn.1 TC0100006494.rn.1 RTA TC
          Rat Expression Set 230 1370506_at - -
          Rat Expression Set 230 1370506_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10725245 - -
          Rat Genome U34 Set rc_AA943150_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22179534 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1369228 - -
          RatRef-12 Expression BeadChip ILMN_53244 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-786bfc498d-47c8f:80/100.66.130.62:80.
          git-commit: 9eb01563f6748445c58f67612c67ecf6b1da7421
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0