Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00682605_g1
          See other DNAJC5 GE Assays ›
          Gene Symbol:
          Dnajc5
          Gene Name:
          DnaJ (Hsp40) homolog, subfamily C, member 5
          Gene Aliases:
          Csp
          Chromosome Location:
          Chr.3: 177013190 - 177047159 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00682605_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Dnajc5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_024161.2 NP_077075.1 4 - 5 616 66
          XM_008762544.1 XP_008760766.1 4 - 5 665 66
          XM_008762545.1 XP_008760767.1 5 - 6 706 66
          GenBank mRNA AF323955.1 - 4 - 5 616 66
          FQ223796.1 - 4 - 5 657 66
          S81917.1 - 3 - 4 516 66
          U39320.1 - 4 - 5 616 66

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300002604.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Dnajc5

          Gene Ontology Categories:

          Function(s) Process(es)

          chemical synaptic transmission
          negative regulation of neuron apoptotic process
          protein binding
          ATP-dependent protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P082042 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 712433 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 251190 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 448315 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 836897 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 821731 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 330987 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 107191 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 603695 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 127967 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 956120 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 392121 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 469238 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 29244 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 699434 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 121786 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 543271 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 430519 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 511866 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 718854 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 323666 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 130751 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 85419 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 24179 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 55989 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P082042 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P082042 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P082042 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P208157 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701937457 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 44519 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 104752 - -
          Agilent Rat cDNA Microarray (G4105A) 701937457 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10843041 - -
          Affymetrix Rat Gene 1.0 ST Array 10843043 - -
          Affymetrix Rat Gene 1.0 ST Array 10843044 - -
          Affymetrix Rat Gene 1.0 ST Array 10843040 - -
          Affymetrix Rat Gene 1.0 ST Array 10843042 - -
          Affymetrix Rat Gene 1.1 ST Array 10843039 - -
          Rat Genome 230 2.0 Array 1371666_at - -
          Rat Genome 230 2.0 Array 1374663_at - -
          Rat Genome 230 2.0 Array 1371666_PM_at - -
          Rat Genome 230 2.0 Array 1369813_PM_at - -
          Rat Genome 230 2.0 Array 1374663_PM_at - -
          Rat Genome 230 2.0 Array 1369813_at - -
          Rat Genome 230 2.0 Array 1384001_PM_at - -
          Rat Genome 230 2.0 Array 1384001_at - -
          Affymetrix HT Rat Focus Array 1369813_PM_at - -
          Affymetrix HT Rat Focus Array 1369813_at - -
          Affymetrix Rat Clariom S Assay PSR0300019303.rn.1 TC0300002604.rn.1 RTA TC
          Rat Expression Set 230 1369813_PM_at - -
          Rat Expression Set 230 1369813_at - -
          Affymetrix Rat Gene 1.0 ST Array 10843039 - -
          Rat Genome U34 Set S81917_s_at - -
          Rat Genome U34 Set U39320_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20857438 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1359259 - -
          RatRef-12 Expression BeadChip ILMN_51895 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0