Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00689010_g1
          See other GSTT2 GE Assays ›
          Gene Symbol:
          Gstt2 LOC103694879
          Gene Name:
          glutathione S-transferase, theta 2
          glutathione S-transferase theta-2-like
          Gene Aliases:
          -
          -
          Chromosome Location:
          Chr.20: 13760810 - 13764481 on Build Rnor_6.0
          Chromosome Location:
          Chr.Un|NW_007906822: 108 - 1485 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Rn00689010_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gstt2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_012796.2 NP_036928.1 4 - 5 661 70
          GenBank mRNA BC061856.1 - 4 - 5 665 70
          CK473092.1 - 4 - 5 661 70
          D10026.1 - 4 - 5 649 70
          FJ179409.1 - 4 - 5 527 70
          Gene Symbol: LOC103694879
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_008773848.1 XP_008772070.1 2 - 3 731 70

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC2000000455.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Gstt2
          Click here to find siRNAs for Gene LOC103694879

          Gene Ontology Categories:

          Function(s) Process(es)

          glutathione metabolic process
          response to salicylic acid
          glutathione transferase activity
          glutathione peroxidase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P547801 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 669633 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 290897 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P547801 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P547801 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P547801 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P194256 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P547801 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 70233 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10829658 - -
          Affymetrix Rat Gene 1.0 ST Array 10829656 - -
          Affymetrix Rat Gene 1.0 ST Array 10829655 - -
          Affymetrix Rat Gene 1.0 ST Array 10829654 - -
          Affymetrix Rat Gene 1.0 ST Array 10829657 - -
          Affymetrix Rat Gene 1.0 ST Array 10829659 - -
          Affymetrix Rat Gene 1.1 ST Array 10829653 - -
          Rat Genome 230 2.0 Array 1368409_PM_at - -
          Rat Genome 230 2.0 Array 1396468_PM_at - -
          Rat Genome 230 2.0 Array 1396468_at - -
          Rat Genome 230 2.0 Array 1368409_at - -
          Affymetrix HT Rat Focus Array 1368409_at - -
          Affymetrix HT Rat Focus Array 1396468_PM_at - -
          Affymetrix HT Rat Focus Array 1368409_PM_at - -
          Affymetrix HT Rat Focus Array 1396468_at - -
          Affymetrix Rat Clariom S Assay PSR2000004444.rn.1 TC2000000455.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR2000004445.rn.1 TC2000000455.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR2000004440.rn.1 TC2000000455.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR2000004442.rn.1 TC2000000455.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR2000004446.rn.1 TC2000000455.rn.1 RTA TC
          Rat Expression Set 230 1368409_PM_at - -
          Rat Expression Set 230 1368409_at - -
          Rat Expression Set 230 1396468_at - -
          Rat Expression Set 230 1396468_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10829653 - -
          Rat Genome U34 Set rc_AI138143_at - -
          Rat Genome U34 Set D10026_s_at - -
          Rat Genome U34 Set D10026_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21796127 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1371280 - -
          RatRef-12 Expression BeadChip ILMN_59083 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0