Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00690866_g1
          See other GUCY1A3 GE Assays ›
          Gene Symbol:
          Gucy1a3
          Gene Name:
          guanylate cyclase 1, soluble, alpha 3
          Gene Aliases:
          Gucy1a1, SGC
          Chromosome Location:
          Chr.2: 181045694 - 181103322 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00690866_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gucy1a3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_017090.2 NP_058786.2 8 - 9 1999 69
          XM_006232518.2 XP_006232580.1 9 - 10 2591 69
          XM_008761107.1 XP_008759329.1 7 - 8 2622 69
          XM_008761108.1 XP_008759330.1 8 - 9 2365 69
          GenBank mRNA AB096020.1 - 7 - 8 1862 69
          BC085746.1 - 8 - 9 1999 69
          M57405.1 - 8 - 9 2318 69
          U60835.1 - 8 - 9 2318 69

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0200003930.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Gucy1a3

          Gene Ontology Categories:

          Function(s) Process(es)

          cGMP biosynthetic process
          nitric oxide mediated signal transduction
          regulation of blood pressure
          response to herbicide
          response to organic cyclic compound
          intracellular signal transduction
          symbiont defense to host-produced nitric oxide
          relaxation of vascular associated smooth muscle
          nucleotide binding
          guanylate cyclase activity
          GTP binding
          heme binding
          ion binding
          protein heterodimerization activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P481167 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P044573 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 765115 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 204900 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 233592 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 515290 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 585428 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 764768 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 417201 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 225646 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 646362 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 562206 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 638433 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 258023 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 220907 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 584979 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 89314 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 316039 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 669952 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 408015 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 663836 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 32947 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 333021 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 189644 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 410765 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 2306 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 883022 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 313749 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 939399 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 432272 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 283859 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 753122 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 436693 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 263222 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 761600 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 734712 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 886500 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 662448 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 922494 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 248023 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 850953 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 960447 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 875758 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 623451 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 55287 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 44185 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 150026 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 144914 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 91160 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 95303 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P481167 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P044573 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P481167 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P044573 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P044573 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P481167 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P320799 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P481167 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 31277 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 99806 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 76035 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 58309 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 80303 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10823921 - -
          Affymetrix Rat Gene 1.0 ST Array 10823931 - -
          Affymetrix Rat Gene 1.0 ST Array 10823925 - -
          Affymetrix Rat Gene 1.0 ST Array 10823923 - -
          Affymetrix Rat Gene 1.0 ST Array 10823930 - -
          Affymetrix Rat Gene 1.0 ST Array 10823924 - -
          Affymetrix Rat Gene 1.0 ST Array 10823929 - -
          Affymetrix Rat Gene 1.0 ST Array 10823922 - -
          Affymetrix Rat Gene 1.0 ST Array 10823928 - -
          Affymetrix Rat Gene 1.0 ST Array 10823926 - -
          Affymetrix Rat Gene 1.0 ST Array 10823927 - -
          Affymetrix Rat Gene 1.1 ST Array 10823920 - -
          Rat Genome 230 2.0 Array 1368154_PM_at - -
          Rat Genome 230 2.0 Array 1368154_at - -
          Rat Genome 230 2.0 Array 1387079_at - -
          Rat Genome 230 2.0 Array 1387079_PM_at - -
          Affymetrix HT Rat Focus Array 1387079_PM_at - -
          Affymetrix HT Rat Focus Array 1368154_PM_at - -
          Affymetrix HT Rat Focus Array 1368154_at - -
          Affymetrix HT Rat Focus Array 1387079_at - -
          Affymetrix Rat Clariom S Assay PSR0200026394.rn.1 TC0200003930.rn.1 RTA TC
          Rat Expression Set 230 1368154_PM_at - -
          Rat Expression Set 230 1368154_at - -
          Rat Expression Set 230 1387079_at - -
          Rat Expression Set 230 1387079_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10823920 - -
          Rat Genome U34 Set rc_AA849036_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21308010 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1371357 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-75d45cbfdf-4zq6t:80/100.66.130.62:80.
          git-commit: a3c66eacce7c9fa3045440ddf4a86215532ee526
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix