Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00692907_m1
          See other SEPP1 GE Assays ›
          Gene Symbol:
          Sepp1
          Gene Name:
          selenoprotein P, plasma, 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.2: 53105912 - 53116198 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00692907_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Sepp1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001083911.1 NP_001077380.1 4 - 5 681 80
          NM_019192.2 NP_062065.2 4 - 5 609 80
          GenBank mRNA BC059137.1 - 4 - 5 610 80
          BC072539.1 - 4 - 5 598 80
          D25221.1 - 4 - 5 590 80
          FQ218512.1 - 2 - 3 238 80
          M63574.1 - 569 80

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0200000478.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Rattus norvegicus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Sepp1

          Gene Ontology Categories:

          Function(s) Process(es)

          selenium compound metabolic process
          brain development
          locomotory behavior
          post-embryonic development
          response to selenium ion
          sexual reproduction
          growth
          selenium binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P057020 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 861333 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 967172 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 607994 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 834580 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 523711 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 297370 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 40319 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 6824 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 452586 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 26713 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P057020 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P057020 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P057020 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P11969 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701880303 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700313711 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 94373 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 95739 - -
          Agilent Rat cDNA Microarray (G4105A) 701880303 - -
          Agilent Rat cDNA Microarray (G4105A) 700313711 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10813242 - -
          Affymetrix Rat Gene 1.0 ST Array 10813238 - -
          Affymetrix Rat Gene 1.0 ST Array 10813240 - -
          Affymetrix Rat Gene 1.0 ST Array 10813243 - -
          Affymetrix Rat Gene 1.0 ST Array 10813241 - -
          Affymetrix Rat Gene 1.0 ST Array 10813239 - -
          Affymetrix Rat Gene 1.0 ST Array 10813237 - -
          Affymetrix Rat Gene 1.1 ST Array 10813236 - -
          Rat Genome 230 2.0 Array 1368806_at - -
          Rat Genome 230 2.0 Array 1368806_PM_at - -
          Rat Genome 230 2.0 Array 1387339_at - -
          Rat Genome 230 2.0 Array 1387339_PM_at - -
          Affymetrix HT Rat Focus Array 1387339_PM_at - -
          Affymetrix HT Rat Focus Array 1387339_at - -
          Affymetrix HT Rat Focus Array 1368806_PM_at - -
          Affymetrix HT Rat Focus Array 1368806_at - -
          Affymetrix Rat Clariom S Assay PSR0200002772.rn.1 TC0200000478.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200002771.rn.1 TC0200000478.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200002773.rn.1 TC0200000478.rn.1 RTA TC
          Rat Expression Set 230 1387339_PM_at - -
          Rat Expression Set 230 1368806_at - -
          Rat Expression Set 230 1387339_at - -
          Rat Expression Set 230 1368806_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10813236 - -
          Rat Genome U34 Set M63574_at - -
          Rat Genome U34 Set rc_AI230247_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21493651 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1649748 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0