Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00694034_m1
          See other LOC102549615 GE Assays ›
          Gene Symbol:
          LOC102549615 Extl3
          Gene Name:
          exostosin-like 3-like
          exostosin-like glycosyltransferase 3
          Gene Aliases:
          -
          Regr
          Chromosome Location:
          Chr.15: 48549363 - 48557235 on Build Rnor_6.0
          Chromosome Location:
          Chr.15: 48420419 - 48465318 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Rn00694034_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC102549615
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_006252223.2 XP_006252285.1 2 - 3 369 73
          Gene Symbol: Extl3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_020097.1 NP_064482.1 4 - 5 2550 73
          XM_006252206.2 XP_006252268.1 7 - 8 3856 73
          GenBank mRNA AB033367.1 - 4 - 5 2550 73

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1500001935.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Rattus norvegicus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene LOC102549615
          Click here to find siRNAs for Gene Extl3

          Gene Ontology Categories:

          Function(s)

          glycosaminoglycan biosynthetic process
          heparan sulfate proteoglycan biosynthetic process
          positive regulation of cell growth

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P161808 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 774044 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 914043 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 888507 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 176898 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 345147 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 41384 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 517728 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 609460 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 428216 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 292484 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 473376 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 908994 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 830604 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 160152 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 158854 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 901004 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 220845 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 88952 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 118654 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 4437 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P161808 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P161808 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P161808 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P311209 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 11217 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 28652 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 61119 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10784541 - -
          Affymetrix Rat Gene 1.0 ST Array 10784544 - -
          Affymetrix Rat Gene 1.0 ST Array 10784543 - -
          Affymetrix Rat Gene 1.0 ST Array 10784545 - -
          Affymetrix Rat Gene 1.0 ST Array 10784542 - -
          Affymetrix Rat Gene 1.1 ST Array 10784540 - -
          Rat Genome 230 2.0 Array 1387489_PM_at - -
          Rat Genome 230 2.0 Array 1387489_at - -
          Affymetrix HT Rat Focus Array 1387489_at - -
          Affymetrix HT Rat Focus Array 1387489_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1500013150.rn.1 TC1500001935.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500013151.rn.1 TC1500001935.rn.1 RTA TC
          Rat Expression Set 230 1387489_PM_at - -
          Rat Expression Set 230 1387489_at - -
          Affymetrix Rat Gene 1.0 ST Array 10784540 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21299849 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1359404 - -
          RatRef-12 Expression BeadChip ILMN_66169 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0