Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00710107_m1
          See other TMEM132A GE Assays ›
          Gene Symbol:
          Tmem132a
          Gene Name:
          transmembrane protein 132A
          Gene Aliases:
          Gbp, Hspa5bp1
          Chromosome Location:
          Chr.1: 226912562 - 226925035 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00710107_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tmem132a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_178021.1 NP_821140.1 4 - 5 853 74
          XM_006231041.2 XP_006231103.1 4 - 5 1788 74
          GenBank mRNA AY216677.1 - 4 - 5 853 74

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100007265.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tmem132a

          Gene Ontology Categories:

          Function(s) Process(es)

          brain development
          negative regulation of programmed cell death
          protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P062545 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 22842 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 349990 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 563823 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 137304 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 62207 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 532135 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 325658 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 454254 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 860235 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 33820 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 23916 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P062545 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P062545 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P062545 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P304011 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 58392 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10728809 - -
          Affymetrix Rat Gene 1.0 ST Array 10728806 - -
          Affymetrix Rat Gene 1.0 ST Array 10728804 - -
          Affymetrix Rat Gene 1.0 ST Array 10728803 - -
          Affymetrix Rat Gene 1.0 ST Array 10728811 - -
          Affymetrix Rat Gene 1.0 ST Array 10728805 - -
          Affymetrix Rat Gene 1.0 ST Array 10728808 - -
          Affymetrix Rat Gene 1.0 ST Array 10728810 - -
          Affymetrix Rat Gene 1.0 ST Array 10728802 - -
          Affymetrix Rat Gene 1.0 ST Array 10728801 - -
          Affymetrix Rat Gene 1.0 ST Array 10728807 - -
          Affymetrix Rat Gene 1.1 ST Array 10728800 - -
          Rat Genome 230 2.0 Array 1383680_at - -
          Rat Genome 230 2.0 Array 1383680_PM_at - -
          Affymetrix HT Rat Focus Array 1383680_at - -
          Affymetrix HT Rat Focus Array 1383680_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0100058816.rn.1 TC0100007265.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100058821.rn.1 TC0100007265.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100058817.rn.1 TC0100007265.rn.1 RTA TC
          Rat Expression Set 230 1383680_PM_at - -
          Rat Expression Set 230 1383680_at - -
          Affymetrix Rat Gene 1.0 ST Array 10728800 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22118665 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6467d75d44-xp7n2:80/100.66.130.62:80.
          git-commit: 8ea1bebb48d33b8b7c0e2103582aa482e6db8664
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0