Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00755416_m1
          See other PRL8A4 GE Assays ›
          Gene Symbol:
          Prl8a4 LOC103694085
          Gene Name:
          prolactin family 8, subfamily a, member 4
          prolactin-8A4
          Gene Aliases:
          Prlph
          -
          Chromosome Location:
          Chr.17: 39376040 - 39382128 on Build Rnor_6.0
          Chromosome Location:
          Chr.17: 39601828 - 39607728 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Rn00755416_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Prl8a4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_021580.1 NP_067591.1 2 - 3 273 107
          XM_008771599.1 XP_008769821.1 2 - 3 209 107
          GenBank mRNA AB009889.1 - 2 - 3 273 107
          Gene Symbol: LOC103694085
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_008771616.1 XP_008769838.1 215 107
          XM_008771617.1 XP_008769839.1 209 107

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1700000594.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Rattus norvegicus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Prl8a4
          Click here to find siRNAs for Gene LOC103694085

          Gene Ontology Categories:

          Process(es)

          hormone activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P164160 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 419812 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 765548 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 664766 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 412559 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 61578 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P164160 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P164160 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P164160 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P287826 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 6730 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10795040 - -
          Affymetrix Rat Gene 1.0 ST Array 10795042 - -
          Affymetrix Rat Gene 1.0 ST Array 10795039 - -
          Affymetrix Rat Gene 1.0 ST Array 10795038 - -
          Affymetrix Rat Gene 1.0 ST Array 10795043 - -
          Affymetrix Rat Gene 1.0 ST Array 10795041 - -
          Affymetrix Rat Gene 1.0 ST Array 10795037 - -
          Affymetrix Rat Gene 1.1 ST Array 10795036 - -
          Rat Genome 230 2.0 Array 1386919_PM_at - -
          Rat Genome 230 2.0 Array 1386920_PM_x_at - -
          Rat Genome 230 2.0 Array 1386919_at - -
          Rat Genome 230 2.0 Array 1386920_x_at - -
          Affymetrix HT Rat Focus Array 1386919_PM_at - -
          Affymetrix HT Rat Focus Array 1386920_x_at - -
          Affymetrix HT Rat Focus Array 1386920_PM_x_at - -
          Affymetrix HT Rat Focus Array 1386919_at - -
          Affymetrix Rat Clariom S Assay PSR1700004100.rn.1 TC1700000594.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700004101.rn.1 TC1700000594.rn.1 RTA TC
          Rat Expression Set 230 1386919_at - -
          Rat Expression Set 230 1386920_PM_x_at - -
          Rat Expression Set 230 1386920_x_at - -
          Rat Expression Set 230 1386919_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10795036 - -
          Rat Genome U34 Set AB009889_f_at - -
          Rat Genome U34 Set rc_AA894246_f_at - -
          Rat Genome U34 Set rc_AI180452_f_at - -
          Rat Genome U34 Set rc_AI180452_i_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1371406 - -
          RatRef-12 Expression BeadChip ILMN_69218 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0