Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00755648_g1
          See other ARF4 GE Assays ›
          Gene Symbol:
          Arf4
          Gene Name:
          ADP-ribosylation factor 4
          Gene Aliases:
          -
          Chromosome Location:
          Chr.16: 2338333 - 2354909 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00755648_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Arf4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_024151.1 NP_077065.1 2 - 3 365 122
          GenBank mRNA BC063167.1 - 2 - 3 323 122
          FQ213125.1 - 2 - 3 348 122
          FQ234083.1 - 2 - 3 323 122
          FQ234248.1 - 2 - 3 435 122
          L12383.1 - 2 - 3 365 122

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1600000034.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Arf4

          Gene Ontology Categories:

          Function(s) Process(es)

          protein ADP-ribosylation
          epidermal growth factor receptor signaling pathway
          small GTPase mediated signal transduction
          brain development
          protein transport
          vesicle-mediated transport
          cell migration
          activation of phospholipase D activity
          negative regulation of apoptotic process
          apical protein localization
          establishment or maintenance of epithelial cell apical/basal polarity
          positive regulation of transcription by RNA polymerase II
          response to axon injury
          protein localization to cilium
          regulation of reactive oxygen species metabolic process
          epidermal growth factor receptor binding
          GTP binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P384223 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 730741 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 563322 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 747972 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 764337 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 797276 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 623770 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 322245 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 746153 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 2288 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 776599 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 508256 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 32494 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 57432 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P384223 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P384223 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P384223 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P384223 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P12406 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700142913 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 84492 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 81506 - -
          Agilent Rat cDNA Microarray (G4105A) 700142913 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10786230 - -
          Affymetrix Rat Gene 1.0 ST Array 10786232 - -
          Affymetrix Rat Gene 1.0 ST Array 10786233 - -
          Affymetrix Rat Gene 1.0 ST Array 10786234 - -
          Affymetrix Rat Gene 1.0 ST Array 10786231 - -
          Affymetrix Rat Gene 1.0 ST Array 10786229 - -
          Affymetrix Rat Gene 1.1 ST Array 10786228 - -
          Rat Genome 230 2.0 Array 1398758_at - -
          Rat Genome 230 2.0 Array 1398758_PM_at - -
          Affymetrix HT Rat Focus Array 1398758_at - -
          Affymetrix HT Rat Focus Array 1398758_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1600000222.rn.1 TC1600000034.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600000225.rn.1 TC1600000034.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600000220.rn.1 TC1600000034.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600000223.rn.1 TC1600000034.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600000224.rn.1 TC1600000034.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600000228.rn.1 TC1600000034.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600000230.rn.1 TC1600000034.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600000221.rn.1 TC1600000034.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600000226.rn.1 TC1600000034.rn.1 RTA TC
          Rat Expression Set 230 1398758_at - -
          Rat Expression Set 230 1398758_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10786228 - -
          Rat Genome U34 Set L12383_at - -
          Rat Genome U34 Set rc_AI028850_f_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22360553 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1358093 - -
          RatRef-12 Expression BeadChip ILMN_63597 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-786bfc498d-47c8f:80/100.66.130.62:80.
          git-commit: 9eb01563f6748445c58f67612c67ecf6b1da7421
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0