Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00769150_g1
          See other RGD1564247 GE Assays ›
          Gene Symbol:
          RGD1564247 LOC103689978 Senp5
          Gene Name:
          similar to SUMO/sentrin specific protease 5
          sentrin-specific protease 5
          SUMO1/sentrin specific peptidase 5
          Gene Aliases:
          -
          -
          -
          Chromosome Location:
          Chr.2: 97694302 - 97697653 on Build Rnor_6.0
          Chromosome Location:
          Chr.11: 72035539 - 72070676 on Build Rnor_6.0
          Chromosome Location:
          Chr.11: 72883891 - 72918756 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          -
          Assay ID Rn00769150_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: RGD1564247
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_003749241.3 XP_003749289.1 1 2358 113
          Gene Symbol: LOC103689978
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_006248475.2 XP_006248537.1 9 - 10 2373 113
          Gene Symbol: Senp5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_221369.8 XP_221369.2 9 - 10 2373 113
          GenBank mRNA FQ233817.1 - 3 - 4 220 113

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Rattus norvegicus Both primers and probe map within a single exon

          Gene Ontology Categories:

          Function(s) Process(es)

          proteolysis
          cysteine-type peptidase activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P076530 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P076530 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P076530 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P277126 - -
          Array Name Probe IDs Targets Target Type
          Rat Genome 230 2.0 Array 1382477_PM_at - -
          Rat Genome 230 2.0 Array 1372594_PM_at - -
          Rat Genome 230 2.0 Array 1391262_at - -
          Rat Genome 230 2.0 Array 1391262_PM_at - -
          Rat Genome 230 2.0 Array 1372594_at - -
          Rat Genome 230 2.0 Array 1382477_at - -
          Affymetrix HT Rat Focus Array 1391262_PM_at - -
          Affymetrix HT Rat Focus Array 1372594_at - -
          Affymetrix HT Rat Focus Array 1372594_PM_at - -
          Affymetrix HT Rat Focus Array 1391262_at - -
          Rat Expression Set 230 1372594_at - -
          Rat Expression Set 230 1372594_PM_at - -
          Rat Expression Set 230 1391262_at - -
          Rat Expression Set 230 1391262_PM_at - -
          Rat Expression Set 230 1382477_at - -
          Rat Expression Set 230 1382477_PM_at - -
          Rat Genome U34 Set rc_AI045059_at - -
          Rat Genome U34 Set rc_AI233721_at - -
          Array Name Probe IDs Targets Target Type
          RatRef-12 Expression BeadChip ILMN_66619 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7699c4d8ff-7jgns:80/100.66.130.62:80.
          git-commit: c11338b6b8d815146c5ef25758b63739976b60cb
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline