Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00788409_s1
          See other TAF9 GE Assays ›
          Gene Symbol:
          Taf9
          Gene Name:
          TAF9 RNA polymerase II, TATA box binding protein (TBP)-associated factor
          Gene Aliases:
          CINAP, TAFII31, TAFII32
          Chromosome Location:
          Chr.2: 30664407 - 30668414 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00788409_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Taf9
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001037310.1 NP_001032387.1 3 733 62
          NM_184048.2 NP_908937.2 3 254 62
          GenBank mRNA AY325235.1 - 2 90 62
          AY359819.1 - 1 50 62
          BC091109.1 - 3 243 62
          CB724309.1 - 3 243 62
          FQ213120.1 - 3 237 62
          FQ215462.1 - 3 248 62
          FQ229736.1 - 3 247 62

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0200000263.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Both primers and probe map within a single exon

          Gene Ontology Categories:

          Function(s) Process(es)

          DNA-templated transcription, initiation
          positive regulation of cell growth
          negative regulation of proteasomal ubiquitin-dependent protein catabolic process
          negative regulation of apoptotic process
          histone H3 acetylation
          positive regulation of transcription by RNA polymerase II
          protein stabilization
          positive regulation of response to cytokine stimulus
          response to interleukin-1
          response to L-glutamate
          transcription cis-regulatory region binding
          p53 binding
          DNA binding
          transcription coactivator activity
          histone acetyltransferase activity
          protein binding
          protein heterodimerization activity
          C2H2 zinc finger domain binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P269125 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P158158 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 358904 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 738235 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 76030 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 246286 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 817063 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 679015 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 971127 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 313059 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 24715 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 785 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P158158 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P269125 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P158158 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P269125 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P158158 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P269125 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P269630 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P269125 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 87029 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 75879 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10812752 - -
          Affymetrix Rat Gene 1.0 ST Array 10812749 - -
          Affymetrix Rat Gene 1.0 ST Array 10812753 - -
          Affymetrix Rat Gene 1.0 ST Array 10832880 - -
          Affymetrix Rat Gene 1.0 ST Array 10812754 - -
          Affymetrix Rat Gene 1.0 ST Array 10812751 - -
          Affymetrix Rat Gene 1.0 ST Array 10812750 - -
          Affymetrix Rat Gene 1.0 ST Array 10940483 - -
          Affymetrix Rat Gene 1.0 ST Array 10812755 - -
          Affymetrix Rat Gene 1.1 ST Array 10832879 - -
          Affymetrix Rat Gene 1.1 ST Array 10940483 - -
          Affymetrix Rat Gene 1.1 ST Array 10812748 - -
          Rat Genome 230 2.0 Array 1375414_PM_at - -
          Rat Genome 230 2.0 Array 1373632_at - -
          Rat Genome 230 2.0 Array 1373632_PM_at - -
          Rat Genome 230 2.0 Array 1375414_at - -
          Affymetrix HT Rat Focus Array 1373632_PM_at - -
          Affymetrix HT Rat Focus Array 1375414_PM_at - -
          Affymetrix HT Rat Focus Array 1375414_at - -
          Affymetrix HT Rat Focus Array 1373632_at - -
          Affymetrix Rat Clariom S Assay PSR0200001482.rn.1 TC0200000263.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200001488.rn.1 TC0200000263.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200001489.rn.1 TC0200000263.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200001480.rn.1 TC0200000263.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200001483.rn.1 TC0200000263.rn.1 RTA TC
          Rat Expression Set 230 1373632_PM_at - -
          Rat Expression Set 230 1375414_at - -
          Rat Expression Set 230 1373632_at - -
          Rat Expression Set 230 1375414_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10812748 - -
          Affymetrix Rat Gene 1.0 ST Array 10832879 - -
          Affymetrix Rat Gene 1.0 ST Array 10940483 - -
          Rat Genome U34 Set rc_AA944572_at - -
          Rat Genome U34 Set rc_AI172041_at - -
          Rat Genome U34 Set rc_AI178819_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20694489 - -
          ABI Rat Genome Survey Microarray 21818340 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1351519 - -
          RatRef-12 Expression BeadChip ILMN_67038 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-97865cc7b-x8sd2:80/100.66.134.75:80.
          git-commit: d53bc399386730d6c5c6d01fe94b21e0ad3fdbfa
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0