Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00820606_m1
          See other PRNP GE Assays ›
          Gene Symbol:
          Prnp
          Gene Name:
          prion protein
          Gene Aliases:
          PrP, Prn
          Chromosome Location:
          Chr.3: 124515922 - 124531320 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00820606_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Prnp
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_012631.2 NP_036763.1 2 - 3 142 60
          GenBank mRNA BC072692.1 - 1 - 2 80 60

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300001759.rn.1

          Additional Information:

          This assay is in the 5'UTR region of the BC072692.1, NM_012631.2 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Prnp

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of protein phosphorylation
          response to oxidative stress
          learning or memory
          negative regulation of interferon-gamma production
          negative regulation of interleukin-17 production
          negative regulation of interleukin-2 production
          regulation of protein localization
          negative regulation of apoptotic process
          negative regulation of DNA-binding transcription factor activity
          negative regulation of activated T cell proliferation
          response to cadmium ion
          response to copper ion
          negative regulation of T cell receptor signaling pathway
          protein homooligomerization
          negative regulation of calcineurin-NFAT signaling cascade
          cellular response to copper ion
          cellular response to xenobiotic stimulus
          regulation of potassium ion transmembrane transport
          copper ion binding
          microtubule binding
          tubulin binding
          identical protein binding
          ATP-dependent protein binding
          chaperone binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P398033 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 787530 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 547044 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 204492 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 819956 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 671657 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 45427 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 886159 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 352657 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 789693 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 213339 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 500373 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 423898 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 75765 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P398033 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P398033 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P398033 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P398033 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 58451 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 19637 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10840078 - -
          Affymetrix Rat Gene 1.0 ST Array 10840077 - -
          Affymetrix Rat Gene 1.0 ST Array 10840079 - -
          Affymetrix Rat Gene 1.1 ST Array 10840076 - -
          Rat Genome 230 2.0 Array 1370156_at - -
          Rat Genome 230 2.0 Array 1370156_PM_at - -
          Affymetrix HT Rat Focus Array 1370156_PM_at - -
          Affymetrix HT Rat Focus Array 1370156_at - -
          Affymetrix Rat Clariom S Assay PSR0300012740.rn.1 TC0300001759.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300012743.rn.1 TC0300001759.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300012739.rn.1 TC0300001759.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300012741.rn.1 TC0300001759.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300012742.rn.1 TC0300001759.rn.1 RTA TC
          Rat Expression Set 230 1370156_at - -
          Rat Expression Set 230 1370156_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10840076 - -
          Rat Genome U34 Set D50093_s_at - -
          Rat Genome U34 Set D50093_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21953535 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376441 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0